Skip to Content
Merck
All Photos(1)

Key Documents

146269

Sigma-Aldrich

4-Chloro-3-methyl-6-nitrophenol

97%

Synonym(s):

4-Chloro-6-nitro-m-cresol

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$350.00

$350.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
25 G
$350.00

About This Item

Linear Formula:
ClC6H2(NO2)(CH3)OH
CAS Number:
Molecular Weight:
187.58
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$350.00


0 left in stock

Request a Bulk Order

Assay

97%

mp

132-134 °C (lit.)

functional group

chloro
nitro

SMILES string

Cc1cc(O)c(cc1Cl)[N+]([O-])=O

InChI

1S/C7H6ClNO3/c1-4-2-7(10)6(9(11)12)3-5(4)8/h2-3,10H,1H3

InChI key

JBMGJOKJUYGIJH-UHFFFAOYSA-N

Application

4-Chloro-3-methyl-6-nitrophenol was used to evaluate toxicity in Tetrahymena pyriformis by quantitative structure-toxicity relationships-topological substructural molecular design method[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maykel Pérez González et al.
Bioorganic & medicinal chemistry, 12(4), 735-744 (2004-02-05)
The TOPological Substructural MOlecular DEsign (TOPS-MODE) has been successfully used in order to explain the toxicity in the Tetrahymena pyriformis on a large data set. The obtained models for the training set had good statistical parameters (R(2)=0.72-0.81, p<0.05) an also

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service