Skip to Content
Merck
All Photos(1)

Key Documents

54306

Supelco

1,4-Dimethoxybenzene

analytical standard

Synonym(s):

p-Methoxyanisole, Dimethylhydroquinone, Hydroquinone dimethyl ether, NSC 7483

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$119.00

$119.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$119.00

About This Item

Linear Formula:
C6H4(OCH3)2
CAS Number:
Molecular Weight:
138.16
Beilstein:
774605
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$119.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

4.8 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥99.0% (GC)

autoignition temp.

1463 °F

shelf life

limited shelf life, expiry date on the label

bp

213 °C (lit.)

mp

54-56 °C (lit.)

density

1.053 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

COc1ccc(OC)cc1

InChI

1S/C8H10O2/c1-9-7-3-5-8(10-2)6-4-7/h3-6H,1-2H3

InChI key

OHBQPCCCRFSCAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

257.0 °F - closed cup

Flash Point(C)

125 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service