Skip to Content
Merck
All Photos(1)

Key Documents

119318

Sigma-Aldrich

Benzene-chromium(0) tricarbonyl

98%

Synonym(s):

Benchrotrene, Benzene-tricarbonylchromium(0)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$161.00

$161.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$161.00

About This Item

Linear Formula:
(C6H6)Cr(CO)3
CAS Number:
Molecular Weight:
214.14
Beilstein:
3571699
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$161.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

reaction suitability

core: chromium
reagent type: catalyst

mp

163-166 °C (lit.)

SMILES string

[Cr].[C-]#[O+].[C-]#[O+].[C-]#[O+].[CH]1[CH][CH][CH][CH][CH]1

InChI

1S/C6H6.3CO.Cr/c1-2-4-6-5-3-1;3*1-2;/h1-6H;;;;

InChI key

WVSBQYMJNMJHIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Oxidation of alkenes
  • Hydrogenation of alkynes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service