Skip to Content
Merck
All Photos(3)

Key Documents

241458

Sigma-Aldrich

Chromium(0) hexacarbonyl

98%

Synonym(s):

Chromium carbonyl, Hexacarbonylchromium

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$220.00
50 G
$840.00

$220.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$220.00
50 G
$840.00

About This Item

Linear Formula:
Cr(CO)6
CAS Number:
Molecular Weight:
220.06
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$220.00


Check Cart for Availability

Request a Bulk Order

vapor density

7.6 (vs air)

vapor pressure

1 mmHg ( 36 °C)

Assay

98%

reaction suitability

core: chromium
reagent type: catalyst

mp

>150 °C (dec.) (lit.)

density

1.77 g/mL at 25 °C (lit.)

SMILES string

[Cr].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]

InChI

1S/6CO.Cr/c6*1-2;

InChI key

KOTQLLUQLXWWDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Precursor to a prenylatedcarbene complex which promotes a net [5+5] cycloaddition of alkynylbenzaldehydes to give phenanthrenes.[1]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suneetha Menon et al.
Tetrahedron, 63(36), 8788-8793 (2008-09-05)
The reaction of prenylated carbene complexes and 2-alkynylbenzoyl derivatives has been investigated. Phenanthrene derivatives are produced if iodine is added prior to product isolation. Under these conditions alkyl migration reactions occur to form the observed products. The product yields are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service