Skip to Content
Merck
All Photos(1)

Key Documents

111937

Sigma-Aldrich

2,4-Dichlorobenzoyl chloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3COCl
CAS Number:
Molecular Weight:
209.46
Beilstein:
608324
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39050538
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.592 (lit.)

bp

150 °C/34 mmHg (lit.)

mp

16-18 °C (lit.)

density

1.494 g/mL at 25 °C (lit.)

functional group

acyl chloride
chloro

SMILES string

ClC(=O)c1ccc(Cl)cc1Cl

InChI

1S/C7H3Cl3O/c8-4-1-2-5(7(10)11)6(9)3-4/h1-3H

InChI key

CEOCVKWBUWKBKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,4-Dichlorobenzoyl chloride was used in the synthesis of biaryls by following the Suzuki coupling[1]. It was used for acylation in the synthesis of potent non-sarcosine-derived Gly T1 inhibitors[2].
2,4-Dichlorobenzoyl chloride was used in the synthesis of thioesters of 4-chlorobenzoate and 2,4-dichlorobenzoate[3].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

278.6 °F - closed cup

Flash Point(C)

137 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Zhijian Zhao et al.
Bioorganic & medicinal chemistry letters, 16(23), 5968-5972 (2006-09-22)
This Letter describes the synthesis and SAR, developed through an iterative analog library approach, of potent and selective non-sarcosine-derived GlyT1 inhibitors.
Markus Thor et al.
Bioorganic & medicinal chemistry letters, 12(24), 3565-3567 (2002-11-22)
A series of 5-substituted 2-benzoylaminobenzoic acids has been synthesized and assayed for PPARalpha/gamma activity. Both dual activators and selective PPARgamma agonists have been identified. This class of compounds was shown to activate the PPARgamma receptor through interaction with a novel
V Romanov et al.
Journal of bacteriology, 178(9), 2656-2661 (1996-05-01)
Corynebacterium sepedonicum KZ-4, described earlier as a strain capable of growth on 2,4-dichlorobenzoate (G.M. Zaitsev and Y.N. Karasevich, Mikrobiologiya 54:356-369, 1985), is known to metabolize this substrate via 4-hydroxybenzoate and protocatechuate, and evidence consistent with an initial reductive dechlorination step

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service