Skip to Content
Merck
All Photos(1)

Key Documents

110043

Sigma-Aldrich

Acetic anhydride

≥99%

Synonym(s):

Acetanhydride, Acetic acid anhydride, Acetyl acetate, Acetyl anhydride, Ethanoic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO)2O
CAS Number:
Molecular Weight:
102.09
Beilstein:
385737
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

3.5 (vs air)

Quality Level

vapor pressure

10 mmHg ( 36 °C)
4 mmHg ( 20 °C)

Assay

≥99%

autoignition temp.

629 °F

expl. lim.

10.3 %

availability

available only in EU

refractive index

n20/D 1.390 (lit.)

bp

138-140 °C (lit.)

mp

−73 °C (lit.)

solubility

water: slightly soluble

density

1.08 g/mL (lit.)

SMILES string

CC(=O)OC(C)=O

InChI

1S/C4H6O3/c1-3(5)7-4(2)6/h1-2H3

InChI key

WFDIJRYMOXRFFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Eagleson M.
Concise Encyclopedia Chemistry, 4-4 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 7-7 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 51-51 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 5-5 (1994)
Lakshmi Pillai-Kastoori et al.
PLoS genetics, 10(7), e1004491-e1004491 (2014-07-11)
Ocular coloboma is a sight-threatening malformation caused by failure of the choroid fissure to close during morphogenesis of the eye, and is frequently associated with additional anomalies, including microphthalmia and cataracts. Although Hedgehog signaling is known to play a critical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service