Skip to Content
Merck
All Photos(1)

Key Documents

105457

Sigma-Aldrich

2,4-Dinitrobenzenesulfenyl chloride

96%

Synonym(s):

2,4-Dinitrophenylsulfenyl chloride, Kharasch reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3SCl
CAS Number:
Molecular Weight:
234.62
Beilstein:
405600
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

mp

94-97 °C (lit.)

solubility

benzene: soluble
diethyl ether: soluble
ethylene chloride: soluble
glacial acetic acid: soluble
methylene chloride: soluble
trichloroethylene: soluble
xylene: soluble

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc(SCl)c(c1)[N+]([O-])=O

InChI

1S/C6H3ClN2O4S/c7-14-6-2-1-4(8(10)11)3-5(6)9(12)13/h1-3H

InChI key

GPXDNWQSQHFKRB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-Dinitrobenzenesulfenyl chloride undergoes addition reactions with transannularly substituted cycloalkenes and norbornenes[1]. 2,4-Dinitrobenzenesulfenyl chloride can convert allylic alcohols to dienes by 1,4-elimination[2].

Application

2,4-Dinitrobenzenesulfenyl chloride was used in preparation of chlorine and bromine biphenyls[2].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of chlorinated and brominated biphenyl oxides.
Reich LL and Reich HJ.
The Journal of Organic Chemistry, 46(18), 3721-3727 (1981)
G K Kumar et al.
Biochemistry, 27(16), 5978-5983 (1988-08-09)
Transcarboxylase (TC) from Propionibacterium shermanii consists of a central hexameric 12S subunit to which 6 outer dimeric 5S subunits are attached through 12 biotinyl 1.3S subunits. The enzyme catalyzes the transfer of a carboxyl group from methylmalonyl-CoA to pyruvate, forming
Studies on the Effect of Remote Substituents on Reactivity: The Rates of Addition of 2, 4-Dinitrobenzenesulfenyl Chloride to Cyclohexenes and Norbornenes (I) 1a.
Kwart H and Miller LJ.
Journal of the American Chemical Society, 83(22), 4552-4559 (1961)
M J De Wolf et al.
The Journal of biological chemistry, 256(11), 5481-5488 (1981-06-10)
The tryptophan residues on cholera toxin and its A and B protomers have been modified by reaction with 2-nitrophenylsulfenyl chloride and 2,4-dinitrophenylsulfenyl chloride. Modification of the tryptophan residues of cholera toxin results in complete loss of toxicity measured in a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service