Skip to Content
Merck
All Photos(2)

Key Documents

206431

Sigma-Aldrich

Dimethyl cyclohexane-1,4-dicarboxylate, mixture of cis and trans

97%

Synonym(s):

1,4-Cyclohexanedicarboxylic dimethyl ester, 1,4-Dimethyl cyclohexane-1,4-dicarboxylate, Dimethyl 1,4-cyclohexanedicarboxylate, Dimethyl hexahydroterephthalate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$179.00
100 G
$383.00

$179.00


Estimated to ship on01 April 2025


Request a Bulk Order

Select a Size

Change View
25 G
$179.00
100 G
$383.00

About This Item

Linear Formula:
C6H10(CO2CH3)2
CAS Number:
Molecular Weight:
200.23
Beilstein:
1876703
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$179.00


Estimated to ship on01 April 2025


Request a Bulk Order

vapor density

6.9 (vs air)

vapor pressure

1 mmHg ( 85 °C)

Assay

97%

autoignition temp.

730 °F

expl. lim.

0.77 %, 150 °F
5.6 %, 184 °F

refractive index

n20/D 1.458 (lit.)

bp

131-133 °C/11 mmHg (lit.)

density

1.111 g/mL at 25 °C

functional group

ester

SMILES string

COC(=O)C1CCC(CC1)C(=O)OC

InChI

1S/C10H16O4/c1-13-9(11)7-3-5-8(6-4-7)10(12)14-2/h7-8H,3-6H2,1-2H3

InChI key

LNGAGQAGYITKCW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dimethyl cyclohexane-1,4-dicarboxylate, mixture of cis and trans may be used in chemical synthesis studies.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service