Skip to Content
Merck
All Photos(1)

Key Documents

102016

Sigma-Aldrich

1,3-Dibromotetrafluorobenzene

96%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$88.90

$88.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$88.90

About This Item

Linear Formula:
Br2C6F4
CAS Number:
Molecular Weight:
307.87
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$88.90


Check Cart for Availability

Request a Bulk Order

Assay

96%

refractive index

n20/D 1.516 (lit.)

bp

200 °C (lit.)

density

2.18 g/mL at 25 °C (lit.)

functional group

bromo
fluoro

SMILES string

Fc1c(F)c(Br)c(F)c(Br)c1F

InChI

1S/C6Br2F4/c7-1-3(9)2(8)5(11)6(12)4(1)10

InChI key

UCWKDDQEZQRGDR-UHFFFAOYSA-N

General description

1,3-Dibromotetrafluorobenzene is a colorless liquid. 1,3-Dibromotetrafluorobenzene crystallizes with 4,4′-dipyridyl to form the noncovalent copolymers[1].

Application

1,3-Dibromotetrafluorobenzene was used to prepare starting compounds for the synthesis of fluorinated liquid crystal derivatives[2]. It was also used in the synthesis of halogen bonded adducts by undergoing In situ cryocrystallization with various oxygenated solvents[3].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

198.0 °F - closed cup

Flash Point(C)

92.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandra De Santis et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 9(16), 3974-3983 (2003-08-14)
The N.Br halogen bonding drives the self-assembly of 1,4-dibromotetrafluorobenzene (1 a) and its 1,3 or 1,2 analogues (1 b,c, respectively) with dipyridyl derivatives 2 a,b. The isomeric supramolecular architectures 3 a-f are obtained as cocrystals that are stable in the
Strategies for the synthesis of fluorinated liquid crystal derivatives from perbromofluoroaromatic systems.
Kenwright AM, et al.
Tetrahedron, 66(52), 9819-9827 (2010)
The Preparation and Reaction of Fluorinated Phenyl Mono-or Bis-Cadmium and Copper Reagents.
Burton DJ, et al.
Journal of Fluorine Chemistry, 52(2), 251-255 (1991)
C?BrO supramolecular synthon: in situ cryocrystallography of low melting halogen-bonded complexes.
CrystEngComm, 14(13), 4259-4261 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service