751413
Di(1-adamantyl)-2-dimethylaminophenylphosphine
98%
Synonym(s):
2-(Di-1-adamantylphosphino)-N,N-dimethylaniline, 2-(Di-1-adamantylphosphino)dimethylaminobenzene, 2-[Bis(tricyclo[3.3.1.13,7]dec-1-yl)phosphino]-N,N-dimethylbenzeneamine, Me-Dalphos
Select a Size
About This Item
Recommended Products
Quality Level
Assay
98%
form
solid
reaction suitability
reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction
mp
237-242 °C
functional group
phosphine
SMILES string
CN(C)c1ccccc1P([C@@]23C[C@@H]4C[C@@H](C[C@@H](C4)C2)C3)[C@@]56C[C@@H]7C[C@@H](C[C@@H](C7)C5)C6
InChI
1S/C28H40NP/c1-29(2)25-5-3-4-6-26(25)30(27-13-19-7-20(14-27)9-21(8-19)15-27)28-16-22-10-23(17-28)12-24(11-22)18-28/h3-6,19-24H,7-18H2,1-2H3/t19-,20+,21-,22-,23+,24-,27-,28-
InChI key
MILNYLCUWRWYBI-XMCXLQRDSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
DalPhos ligands enable Pd-catalyzed C-N and C-C bond formation with good functional group tolerance.
Related Content
Stradiotto group focuses on electronically rich ligands for late metal catalysis, including Pd and Au chemistry applications.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service