Skip to Content
Merck
All Photos(2)

Key Documents

910732

Sigma-Aldrich

CyPAd-DalPhos

≥95%

Synonym(s):

8-(2-(Dicyclohexylphosphaneyl)phenyl)-1,3,5,7-tetramethyl-2,4,6-trioxa-8-phosphaadamantane

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$563.00

$563.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$563.00

About This Item

Empirical Formula (Hill Notation):
C28H42O3P2
CAS Number:
Molecular Weight:
488.58
MDL number:
UNSPSC Code:
12352112
NACRES:
NA.22

$563.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95%

form

powder

reaction suitability

reagent type: ligand

mp

181-182 °C

functional group

phosphine

SMILES string

P4(C5(OC6(OC(OC4(C6)C)(C5)C)C)C)c1c(cccc1)P(C3CCCCC3)C2CCCCC2

InChI

1S/C28H42O3P2/c1-25-19-27(3)31-26(2,29-25)20-28(4,30-25)33(27)24-18-12-11-17-23(24)32(21-13-7-5-8-14-21)22-15-9-6-10-16-22/h11-12,17-18,21-22H,5-10,13-16,19-20H2,1-4H3

InChI key

RDFYIHPJEXXODP-UHFFFAOYSA-N

Related Categories

Application

CyPAd-DalPhos is ligand developed in the Stradiotto lab that when combined with a Ni(II) source forms an air stable pre catalyst for C(sp2)-O coupling and N-arylation reactions.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nickel-Catalyzed N-Arylation of Cyclopropylamine and Related Ammonium Salts with (Hetero)aryl (Pseudo)halides at Room Temperature
Tassone J P, et al
ACS Catalysis, 7(9), 6048-6059 (2017)
Christopher M Lavoie et al.
Nature communications, 7, 11073-11073 (2016-03-24)
Palladium-catalysed C(sp(2))-N cross-coupling (that is, Buchwald-Hartwig amination) is employed widely in synthetic chemistry, including in the pharmaceutical industry, for the synthesis of (hetero)aniline derivatives. However, the cost and relative scarcity of palladium provides motivation for the development of alternative, more

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service