Merck
  • Quantification of volatile-alkylated selenium and sulfur in complex aqueous media using solid-phase microextraction.
Quantification of volatile-alkylated selenium and sulfur in complex aqueous media using solid-phase microextraction.
Journal of chromatography. A (2015-07-08)
Bas Vriens, Marcel Mathis, Lenny H E Winkel, Michael Berg
ABSTRACT
Biologically produced volatile-alkylated Se and S compounds play an important role in the global biogeochemical Se and S cycles, are important constituents of odorous industrial emissions, and contribute to (off-)flavors in food and beverages. This study presents a fully automated direct-immersion solid-phase microextraction (DI-SPME) method coupled with capillary gas chromatography-mass spectrometry (GC/MS) for the simultaneous quantification of 10 volatile-alkylated Se and S compounds in complex aqueous media. Instrumental parameters of the SPME procedure were optimized to yield extraction efficiencies of up to 96% from complex aqueous matrices. The effects of sample matrix composition and analyte transformation during sample storage were critically assessed. With the use of internal standards and procedural calibrations, the DI-SPME-GC/MS method allows for trace-level quantification of volatile Se and S compounds in the ng/L range (e.g. down to 30 ng/L dimethyl sulfide and 75 ng/L dimethyl selenide). The applicability and robustness of the presented method demonstrate that the method may be used to quantify volatile Se and S compounds in complex aqueous samples, such as industrial effluents or food and beverage samples.
Product Description
Supelco
SPME StableFlex fiber assortment kit, for use with manual holder
Supelco
SPME fiber assortment kit 1, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assortment kit 2, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assortment kit 4, for use with autosampler, size 24 ga
Supelco
SPME StableFlex fiber assortment kit, needle size 23 ga, StableFlex fiber, for use with autosampler
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 23 ga
Supelco
SPME StableFlex fiber assortment kit, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assortment kit 2, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assortment kit 4, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assortment kit 2, for use with autosampler, size 24 ga
Supelco
SPME fiber assortment kit 4, for use with manual holder, size 24 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, for use with autosampler
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga, metal alloy fiber
Supelco
SPME fiber assortment kit 5, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm(PEG, for use with autosampler
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm(PEG, metal alloy fiber, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), metal alloy fiber, df 100 μm(PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 23 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 23 ga, PDMS/DVB StableFlex, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 23 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), fused silica fiber, df 65 μm(PDMS/DVB, for use with manual holder, needle size 23 ga