Merck
  • Elucidating toxicological mechanisms of current flame retardants using a bacterial gene profiling assay.
Elucidating toxicological mechanisms of current flame retardants using a bacterial gene profiling assay.
Toxicology in vitro : an international journal published in association with BIBRA (2015-09-08)
Boris V Krivoshiev, Freddy Dardenne, Ronny Blust, Adrian Covaci, Steven J Husson
ABSTRACT
Flame retardants are ubiquitously used chemicals that have been shown to contaminate environments. Toxicological data is largely limited, with little insight into their molecular modes of action that may give rise to their toxic phenotypes. Such insight would aid more effective risk assessments concerning these compounds, while also improving molecular design. We therefore used a bacterial stress-gene profiling assay to screen twelve currently-used flame retardants to obtain mechanistic insights of toxicity. Both brominated and organophosphate flame retardants were tested. All compounds showed statistically significant inductions of several stress genes when compared to control treatments. Triphenyl phosphate, tris(2-butoxyethyl) phosphate, tris(1,3-dichloro-2-propyl)phosphate, tris(butyl)phosphate, and tetrabromobisphenol A elicited (at least) two-fold inductions for any of the stress genes. When looking at absolute induction levels, the promoters induced are indicative of protein perturbation, DNA integrity and membrane integrity. However, normalising for the different induction potentials of the different stress genes and clustering using hierarchical and k-means algorithms indicated that in addition to protein and DNA damage, some compounds also resulted in growth arrest and oxidative damage. This research shows that this assay allows for the determination of toxicological modes-of-action while clustering and accounting for induction potentials of the different genes aids better risk assessment.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Triphenyl phosphate solution, suitable for NMR (reference standard), 0.0485 M in acetone-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
3,3′,5,5′-Tetrabromobisphenol A, 97%
Sigma-Aldrich
Triphenyl phosphate solution, suitable for NMR (reference standard), 0.0485 M in acetone-d6 (99.9 atom % D)
Sigma-Aldrich
Triphenyl phosphate solution, suitable for NMR (reference standard), 0.0485 M in acetone-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Triethyl phosphate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Tributyl phosphate, ≥99%
Sigma-Aldrich
Tributyl phosphate, 97%
Sigma-Aldrich
Triphenyl phosphate, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Supelco
Triphenyl phosphate, analytical standard
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Tetrabutylphosphonium hydroxide solution, 40 wt. % in H2O
Sigma-Aldrich
Tris(2-butoxyethyl) phosphate, 94%
Sigma-Aldrich
Tris(2-chloroethyl) phosphate, 97%