Skip to Content
Merck
All Photos(1)

Key Documents

733741

Sigma-Aldrich

Triphenyl phosphate solution

NMR reference standard, 0.0485 M in acetone-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€387.00

€387.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 PKG
€387.00

About This Item

Linear Formula:
(C6H5O)3PO
Molecular Weight:
326.28
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.12

€387.00


Check Cart for Availability

Request more information

grade

NMR reference standard

form

solution

concentration

0.0485 M in acetone-d6 (99.9 atom % D)

technique(s)

NMR: suitable

NMR tube size

3 mm × 8 in.

suitability

suitable for NMR (reference standard)

SMILES string

O=P(Oc1ccccc1)(Oc2ccccc2)Oc3ccccc3

InChI

1S/C18H15O4P/c19-23(20-16-10-4-1-5-11-16,21-17-12-6-2-7-13-17)22-18-14-8-3-9-15-18/h1-15H

InChI key

XZZNDPSIHUTMOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Features and Benefits

31P sensitivity

Quantity

3 mm O.D. tube contains 0.235 mL.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-2.2 °F

Flash Point(C)

-19 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sin-Ying Koou et al.
Parasites & vectors, 7, 471-471 (2014-10-11)
In Singapore, dose-response bioassays of Aedes aegypti (L.) adults have been conducted, but the mechanisms underlying resistance to insecticides remain unclear. In this study, we evaluated insecticide resistance and its underlying mechanism in field populations of Ae. aegypti adults. Seven
M I Cervera et al.
Analytical and bioanalytical chemistry, 406(27), 6843-6855 (2014-05-16)
An atmospheric pressure chemical ionization source has been used to enhance the potential of gas chromatography coupled with quadrupole time-of-flight (QTOF) mass spectrometry (MS) for screening and quantification purposes in pesticide residue analysis. A screening method developed in our laboratory
Francesc A Esteve-Turrillas et al.
PloS one, 10(7), e0134042-e0134042 (2015-07-28)
Bioconjugate preparation is a fundamental step for antibody generation and immunoassay development to small chemical compounds. For analytical targets holding in their structure an aryl halogen atom, cross-coupling reactions may be a simple and efficient way to obtain functionalized derivatives;
Davide Savy et al.
Molecules (Basel, Switzerland), 20(11), 19958-19970 (2015-11-12)
The molecular composition of water-soluble lignins isolated from four non-food bioenergy crops (cardoon CAR, eucalyptus EUC, and two black poplars RIP and LIM) was characterized in detail, and their potential bioactivity towards maize germination and early growth evaluated. Lignins were
Boris V Krivoshiev et al.
Toxicology in vitro : an international journal published in association with BIBRA, 29(8), 2124-2132 (2015-09-08)
Flame retardants are ubiquitously used chemicals that have been shown to contaminate environments. Toxicological data is largely limited, with little insight into their molecular modes of action that may give rise to their toxic phenotypes. Such insight would aid more

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service