Skip to Content
Merck
All Photos(1)

Documents

673439

Sigma-Aldrich

Dimethyl sulfoxide solution

50 wt. % in H2O

Synonym(s):

DMSO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H6OS
CAS Number:
Molecular Weight:
78.13
MDL number:
UNSPSC Code:
12191600
PubChem Substance ID:

form

liquid

Quality Level

concentration

50 wt. % in H2O

density

1.069 g/mL at 25 °C

SMILES string

CS(C)=O

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

InChI key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Schlam et al.
Nature communications, 6, 8623-8623 (2015-10-16)
Phagocytosis is responsible for the elimination of particles of widely disparate sizes, from large fungi or effete cells to small bacteria. Though superficially similar, the molecular mechanisms involved differ: engulfment of large targets requires phosphoinositide 3-kinase (PI3K), while that of
Mike Clements et al.
Toxicological sciences : an official journal of the Society of Toxicology, 140(2), 445-461 (2014-05-09)
Human stem cell derived cardiomyocytes (hESC-CM) provide a potential model for development of improved assays for pre-clinical predictive drug safety screening. We have used multi-electrode array (MEA) analysis of hESC-CM to generate multi-parameter data to profile drug impact on cardiomyocyte
D B Bas et al.
European journal of pain (London, England), 19(2), 260-270 (2014-06-20)
Mounting evidence points to individual contributions of tumour necrosis factor-alpha (TNF) and the c-Jun N-terminal kinase (JNK) pathway to the induction and maintenance of various pain states. Here we explore the role of spinal TNF and JNK in carrageenan-induced hypersensitivity.
Xin Yao et al.
PloS one, 9(7), e101564-e101564 (2014-07-09)
The mitochondrial Bit1 (Bcl-2 inhibitor of transcription 1) protein is a part of an apoptotic pathway that is uniquely regulated by integrin-mediated attachment. As an anoikis effector, Bit1 is released into the cytoplasm following loss of cell attachment and induces
C Mathieu et al.
Journal of virology, 89(2), 1143-1155 (2014-11-08)
Measles virus (MV) infection is undergoing resurgence and remains one of the leading causes of death among young children worldwide despite the availability of an effective measles vaccine. MV infects its target cells by coordinated action of the MV H

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service