Merck
  • Quantitative analysis of phenylalanine, tyrosine, tryptophan and kynurenine in rat model for tauopathies by ultra-high performance liquid chromatography with fluorescence and mass spectrometry detection.
Quantitative analysis of phenylalanine, tyrosine, tryptophan and kynurenine in rat model for tauopathies by ultra-high performance liquid chromatography with fluorescence and mass spectrometry detection.
Journal of pharmaceutical and biomedical analysis (2015-09-08)
Jaroslav Galba, Alena Michalicova, Vojtech Parrak, Michal Novak, Andrej Kovac
ABSTRACT
We developed and validated a simple and sensitive ultra-high performance liquid chromatography (UHPLC) method for the analysis of phenylalanine (Phe), tyrosine (Tyr), tryptophan (Trp) and kynurenine (Kyn) in rat plasma. Analytes were separated on Acquity UPLC HSS T3 column (2.1 mm×50 mm, 1.8 μm particle size) using a 4 min ammonium acetate (pH 5) gradient and detected by fluorescence and positive ESI mass spectrometry. Sample preparation involved dilution of plasma, deproteinization by trichloroacetic acid and centrifugation. The procedure was validated in compliance with the FDA guideline. The limits of quantification (LOQ) were 0.3 μM for Kyn and from 1.5 to 3 μM for Phe, Tyr, Trp. The method showed excellent linearity with regression coefficients higher than 0.99. The accuracy was within the range of 86-108%. The inter-day precision (n=5 days), expressed as % RSD, was in the range 1-13%. The benefit of using UHPLC is a short analysis period and thus, a very good sample throughput. Using this method, we analyzed plasma samples and detected significant changes of Kyn and Phe in transgenic rat model for tauopathies.
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
SAFC
L-Tyrosine
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
L-Phenylalanine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
L-Phenylalanine, 99%, FCC