Skip to Content
Merck
All Photos(1)

Key Documents

10300

Sigma-Aldrich

5α-Androstan-17β-ol-3-one

purum, ≥99.0% (TLC)

Synonym(s):

17β-Hydroxy-5α-androstan-3-one, 4,5α-Dihydrotestosterone, Androstanolone, Stanolone

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€45.30
5 G
€179.00

€45.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€45.30
5 G
€179.00

About This Item

Empirical Formula (Hill Notation):
C19H30O2
CAS Number:
Molecular Weight:
290.44
Beilstein:
2056371
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

€45.30


Check Cart for Availability

Request a Bulk Order

biological source

synthetic

Quality Level

sterility

non-sterile

grade

purum

Assay

≥99.0% (TLC)

form

powder

optical activity

[α]20/D +33±1°, c = 1% in ethanol

drug control

USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada

mp

178-183 °C

storage temp.

2-8°C

SMILES string

[H][C@@]12CC[C@@]3([H])[C@]4([H])CC[C@H](O)[C@@]4(C)CC[C@]3([H])[C@@]1(C)CCC(=O)C2

InChI

1S/C19H30O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h12,14-17,21H,3-11H2,1-2H3/t12-,14-,15-,16-,17-,18-,19-/m0/s1

InChI key

NVKAWKQGWWIWPM-ABEVXSGRSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Sales restrictions may apply

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bu B Yeap et al.
The Journal of clinical endocrinology and metabolism, 99(1), E9-18 (2013-11-22)
Testosterone (T) levels decline with age and lower T has been associated with increased mortality in aging men. However, the associations of its metabolites, dihydrotestosterone (DHT) and estradiol (E2), with mortality are poorly defined. We assessed associations of T, DHT
Takashi Taniguchi et al.
Arthritis & rheumatology (Hoboken, N.J.), 67(2), 517-526 (2014-11-12)
Fli-1, a potential predisposing factor for systemic sclerosis (SSc), is constitutively down-regulated in the lesional skin of patients with SSc by an epigenetic mechanism. To investigate the impact of Fli-1 deficiency on the induction of an SSc phenotype in various
Isabelle Brunet et al.
The Journal of clinical investigation, 124(7), 3230-3240 (2014-06-18)
Autonomic sympathetic nerves innervate peripheral resistance arteries, thereby regulating vascular tone and controlling blood supply to organs. Despite the fundamental importance of blood flow control, how sympathetic arterial innervation develops remains largely unknown. Here, we identified the axon guidance cue
Katharine Trenholme et al.
Antimicrobial agents and chemotherapy, 58(7), 3666-3678 (2014-04-16)
Therapies to prevent transmission of malaria parasites to the mosquito vector are a vital part of the global malaria elimination agenda. Primaquine is currently the only drug with such activity; however, its use is limited by side effects. The development
Ken Ye et al.
PloS one, 9(6), e99410-e99410 (2014-06-12)
Infrapatellar fat pad adipose stem cells (IPFP-ASCs) have been shown to harbor chondrogenic potential. When combined with 3D polymeric structures, the stem cells provide a source of stem cells to engineer 3D tissues for cartilage repair. In this study, we

Questions

  1. 細胞培養に使用できますか?その際の使用は培地に直接粉末で添加するのでよいのでしょうか。

    1 answer
    1. This product has not been sterilized, and it is not recommended to add non-sterile powder directly to cell culture medium due to the risk of introducing contaminants. It is highly recommended to filter-sterilize the reconstituted solution of this product before applying it to cell culture.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service