Skip to Content
Merck
All Photos(5)

Key Documents

T3754

Sigma-Aldrich

L-Tyrosine

≥98% (HPLC)

Synonym(s):

(S)-2-Amino-3-(4-hydroxyphenyl)propionic acid, 3-(4-Hydroxyphenyl)-L-alanine

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€49.70
100 G
€80.10
500 G
€331.00
1 KG
€570.00

€49.70


Estimated to ship on26 March 2025


Request a Bulk Order

Select a Size

Change View
50 G
€49.70
100 G
€80.10
500 G
€331.00
1 KG
€570.00

About This Item

Linear Formula:
4-(HO)C6H4CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
181.19
Beilstein:
392441
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

€49.70


Estimated to ship on26 March 2025


Request a Bulk Order

Product Name

L-Tyrosine, reagent grade, ≥98% (HPLC)

grade

reagent grade

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

mp

>300 °C (dec.) (lit.)

solubility

1 M HCl: soluble 50 mg/mL

application(s)

detection

SMILES string

N[C@@H](Cc1ccc(O)cc1)C(O)=O

InChI

1S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11H,5,10H2,(H,12,13)/t8-/m0/s1

InChI key

OUYCCCASQSFEME-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Tyrosine has been used to prepare tyrosine solution for tyrosinase assay. It has also been used to assay pepsin activity.[1][2]

Biochem/physiol Actions

L-Tyrosine is a non-essential amino acid with a polar side chain. It is utilized by cells to synthesize proteins that have a role in signal transduction.[3] L-Tyrosine is a proteogenic amino acid that acts as a receiver of phosphate groups that are transferred by kinases.[4]

Other Notes

Amino acid precursor of dopamine and other catecholamines

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karen Timberlake
Chemistry, An Introduction to General, Organic, and Biological Chemistry (2016)
Effect of insoluble fiber supplementation applied at different ages on digestive organ weight and digestive enzymes of layer-strain poultry
J. S. Yokhana
Poultry Science, 95(3), 550-559 (2016)
Study of Physiochemical Parameters for Optimizing Tyrosinase Activity in Rhodococcus ruber C4 and Pseudomonas geniculata C7/C10
Sapna S. Ingle
Journal of Pharmacy Research, 9(6) (2015)
John W Hill
Chemistry for Changing Times (2016)
Hideki Hara et al.
Nature immunology, 14(12), 1247-1255 (2013-11-05)
The inflammasome adaptor ASC contributes to innate immunity through the activation of caspase-1. Here we found that signaling pathways dependent on the kinases Syk and Jnk were required for the activation of caspase-1 via the ASC-dependent inflammasomes NLRP3 and AIM2.

Protocols

Proteinase K activity measured via spectrophotometry using hemoglobin substrate, crucial for enzyme characterization.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service