Merck
Chemical analysis of Karenia papilionacea.
Toxicon : official journal of the International Society on Toxinology (2015-05-20)
Nicholas Fowler, Carmelo Tomas, Daniel Baden, Lisa Campbell, Andrea Bourdelais
ABSTRACT
One of the most widely studied organisms responsible for Harmful Algal Blooms (HABs) is the marine dinoflagellate Karenia brevis. This organism produces neurotoxic compounds known as brevetoxins. A related dinoflagellate, Karenia papilionacea, has been reported to occasionally co-bloom with K. brevis but has received little attention as a possible toxin producing species. Therefore, our aim was to investigate the toxin profile for K. papilionacea. A toxic fraction was identified using a cell based cytotoxicity assay and the toxin was isolated and identified as the ladder frame polyether brevetoxin-2 (PbTx-2) using mass spectrometry (MS) and nuclear magnetic resonance (NMR). Toxin production in K. papilionacea increased in response to hypoosmotic stress, as previously observed in K. brevis.
Sigma-Aldrich
Manganese(II) chloride, powder and chunks, ≥99% trace metals basis
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Manganese(II) chloride, beads, 98%
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Manganese(II) chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Manganese(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Cobalt(II) chloride, AnhydroBeads, −10 mesh, 99.995% trace metals basis
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Methanol-d4, "100%", ≥99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Cobalt(II) chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Methanol-d4, anhydrous, ≥99.8 atom % D
Sigma-Aldrich
Thiamine hydrochloride, ≥98%, FCC, FG
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
Methanol-d4, "100%", 99.96 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, 99 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture