Skip to Content
Merck

Skip To

S5022

Sodium nitrate

≥99.0%, suitable for plant cell culture

Synonym(s):

Chile salpeter

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kg
Check Cart for Availability
€ 116,00

About This Item

Linear Formula:
NaNO3
CAS Number:
Molecular Weight:
84.99
NACRES:
NA.71
PubChem Substance ID:
UNSPSC Code:
12352207
EC Number:
231-554-3
MDL number:
Assay:
≥99.0%
Solubility:
water: soluble 2 g/10 mL, colorless

€ 116,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99.0%

technique(s)

cell culture | plant: suitable

mp

306 °C (dec.) (lit.)

solubility

water: soluble 2 g/10 mL, colorless

SMILES string

[Na+].[O-][N+]([O-])=O

InChI

1S/NO3.Na/c2-1(3)4;/q-1;+1

InChI key

VWDWKYIASSYTQR-UHFFFAOYSA-N

Application


  • Sodium salt promoted the generation of nano zero valent iron by carbothermal reduction: For activating peroxydisulfate to degrade antibiotic.: This study investigates the role of sodium nitrate in enhancing the generation of nano zero-valent iron, a critical component for activating peroxydisulfate in antibiotic degradation processes, showcasing its potential in environmental remediation technologies (Xue C et al., 2024).

  • Fabricating Precise and Smooth Microgroove Structures on Zr-Based Metallic Glass Using Jet-ECM.: Although indirectly related, the inclusion of sodium nitrate in the electrolyte solution for jet electrochemical machining (ECM) demonstrates its utility in improving manufacturing processes for precision engineering (Li D et al., 2024).


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
221341S817071755
assay

≥99.0%

assay

≥99.0%

assay

≥99.0%

assay

≥99.0% (T)

solubility

water: soluble 2 g/10 mL, colorless

solubility

-

solubility

H2O: 1 M at 20 °C, clear, colorless

solubility

H2O: 1 M at 20 °C, clear, colorless

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200

mp

306 °C (dec.) (lit.)

mp

306 °C (dec.) (lit.)

mp

306 °C (dec.) (lit.)

mp

306 °C (dec.) (lit.)

technique(s)

cell culture | plant: suitable

technique(s)

-

technique(s)

-

technique(s)

-


Still not finding the right product?

Explore all of our products under Sodium nitrate


pictograms

Flame over circleExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class

5.1B - Oxidizing hazardous materials

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters