Merck
  • Rapid screening of edible oils for phthalates using phase-transfer catalyst-assisted hydrolysis and liquid phase microextraction coupled to high performance liquid chromatography-tandem mass spectrometry.
Rapid screening of edible oils for phthalates using phase-transfer catalyst-assisted hydrolysis and liquid phase microextraction coupled to high performance liquid chromatography-tandem mass spectrometry.
Journal of chromatography. A (2015-10-20)
Shuhui Liu, Laping Liu, Yangying Han, Jingru Sun, Jing Feng, Jin Wang, Chongming Zhong
ABSTRACT
Edible oil is easily contaminated with phthalic acid esters (PAEs). Conventional procedures to analyze individual PAEs require very rigorous experimental conditions that are extremely labor-intensive due to significant procedural contaminations generated by the ubiquitous presence of PAEs in the laboratory environment. In this study, a rapid screening method for PAEs in edible oil was successfully developed. Using a phase-transfer catalyst (terabutylammonium bromide) during oil/water biphasic base hydrolysis of PAEs, the hydrolysis time was decreased from a previously reported time of 20 h to 10 min (80 °C). The resulting phthalic acid in the acidified hydrolysate was extracted with 600 μL of tributyl phosphate and then analyzed by high performance liquid chromatography-tandem mass spectrometry in 6 min. Parameters affecting the hydrolysis of PAEs and the extraction of phthalic acid were optimized, and the analytical method was validated. No obvious matrix effect existed in the edible oils whether an external or internal standard method was used. The detection limit was 1.0 μmol kg(-1), and the quantification limit was 1.3 μmol kg(-1). The recovery rates varied from 86 to 107% with relative standard deviations equal to or lower than 9.9% in all of the tested conditions. Twenty-six samples were analyzed, and the background corrected total PAE content was found to be in the range of <LOD-52.1 μmol kg(-1). This fast and reliable method was not only a practical way to screen oil samples but can also be used as an indicator of false positive or overestimated results in conventional analysis of PAEs. In addition, it presents a new and promising methodology to deal with oil matrices.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Dess-Martin periodinane, 97%
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
1-Octanol, ReagentPlus®, 99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Tetrabutylammonium bromide, ACS reagent, ≥98.0%
Sigma-Aldrich
Diethyl carbonate, anhydrous, ≥99%
Sigma-Aldrich
Tributyl phosphate, 97%
Sigma-Aldrich
Tetrabutylammonium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Tributylphosphine solution, 200 mM (in N-methyl-2-pyrrolidinone), liquid
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Tetrabutylammonium chloride, ≥97.0% (NT)
Sigma-Aldrich
Diethyl carbonate, 99%
Sigma-Aldrich
Tributyl phosphate, ≥99%
Sigma-Aldrich
Tetrabutylammonium bromide solution, 50 wt. % in H2O
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%