Merck
  • The N-ethylmaleimide-sensitive factor and dysbindin interact to modulate synaptic plasticity.
The N-ethylmaleimide-sensitive factor and dysbindin interact to modulate synaptic plasticity.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2015-05-15)
Avanti Gokhale, Ariana P Mullin, Stephanie A Zlatic, Charles A Easley, Megan E Merritt, Nisha Raj, Jennifer Larimore, David E Gordon, Andrew A Peden, Subhabrata Sanyal, Victor Faundez
ABSTRACT
Dysbindin is a schizophrenia susceptibility factor and subunit of the biogenesis of lysosome-related organelles complex 1 (BLOC-1) required for lysosome-related organelle biogenesis, and in neurons, synaptic vesicle assembly, neurotransmission, and plasticity. Protein networks, or interactomes, downstream of dysbindin/BLOC-1 remain partially explored despite their potential to illuminate neurodevelopmental disorder mechanisms. Here, we conducted a proteome-wide search for polypeptides whose cellular content is sensitive to dysbindin/BLOC-1 loss of function. We identified components of the vesicle fusion machinery as factors downregulated in dysbindin/BLOC-1 deficiency in neuroectodermal cells and iPSC-derived human neurons, among them the N-ethylmaleimide-sensitive factor (NSF). Human dysbindin/BLOC-1 coprecipitates with NSF and vice versa, and both proteins colocalized in a Drosophila model synapse. To test the hypothesis that NSF and dysbindin/BLOC-1 participate in a pathway-regulating synaptic function, we examined the role for NSF in dysbindin/BLOC-1-dependent synaptic homeostatic plasticity in Drosophila. As previously described, we found that mutations in dysbindin precluded homeostatic synaptic plasticity elicited by acute blockage of postsynaptic receptors. This dysbindin mutant phenotype is fully rescued by presynaptic expression of either dysbindin or Drosophila NSF. However, neither reduction of NSF alone or in combination with dysbindin haploinsufficiency impaired homeostatic synaptic plasticity. Our results demonstrate that dysbindin/BLOC-1 expression defects result in altered cellular content of proteins of the vesicle fusion apparatus and therefore influence synaptic plasticity.
Sigma-Aldrich
Anti-DTNBP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab3
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanesulfonic acid, 95%
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Pricing and availability is not currently available.
SAFC
BIS-TRIS
Pricing and availability is not currently available.
SAFC
HEPES
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium chloride solution, 1.00 M±0.01 M
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-MAP2 (2a+2b) antibody, Mouse monoclonal, clone AP-20, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 5 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, tablet
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, DNase, RNase, and protease, none detected, ≥99% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Pricing and availability is not currently available.