Merck
Cyclophilin-D: a resident regulator of mitochondrial gene expression.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2015-04-04)
Jeejabai Radhakrishnan, Stanley Bazarek, Bala Chandran, Raúl J Gazmuri
ABSTRACT
Cyclophilin-D (Cyp-D) is a mitochondrial matrix peptidyl-prolyl isomerase. Because cyclophilins can regulate nuclear gene expression, we examined whether Cyp-D could regulate mitochondrial gene expression. We demonstrated in HEK 293T cells that transfected Cyp-D interacts with mitochondrial transcription factors B1 and B2 (TFB2M) but not with mitochondrial transcription factor A. We also demonstrated that Cyp-D interacts in vivo with TFB2M. Genetic silencing of Cyp-D and pharmacologic inhibition of Cyp-D markedly reduced mitochondrial transcription to 18 ± 5% (P < 0.05) and 24 ± 3% (P < 0.05) of respective controls. The level of interaction between Cyp-D and TFB2M correlated with the level of nascent mitochondrial RNA intensity (r = 0.896; P = 0.0156). Cyp-D silencing down-regulated mitochondrial transcripts initiated from the heavy strand promoter 2 [i.e., NADH dehydrogenase 1 (ND1) by 11-fold, P < 0.005; cytochrome oxidase 1 (COX1) by 4-fold, P < 0.001; and ATP synthase subunit 6 (ATP6) by 6.5-fold, P < 0.005); but not NADH dehydrogenase 6 (ND6)], which is initiated from the light strand promoter. Cyp-D silencing reduced mitochondrial membrane potential and cellular oxygen consumption (from 59 ± 5 to 34 ± 1 µmol oxygen/min/10(6) cells, P < 0.001); the latter without a statistically significant reversal after uncoupling electron transport from ATP synthesis, consistent with down-regulation of electron transport complexes. Accordingly, these studies provide novel evidence that Cyp-D could play a key role in regulating mitochondrial gene expression.
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Tetramethylrhodamine ethyl ester perchlorate, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
SAFC
BIS-TRIS
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium pyrophosphate tetrabasic, ≥95%
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis