Merck
  • Required role of apoptotic myogenic precursors and toll-like receptor stimulation for the establishment of autoimmune myositis in experimental murine models.
Required role of apoptotic myogenic precursors and toll-like receptor stimulation for the establishment of autoimmune myositis in experimental murine models.
Arthritis & rheumatology (Hoboken, N.J.) (2014-12-17)
Clara Sciorati, Antonella Monno, Dana P Ascherman, Elena Seletti, Angelo A Manfredi, Patrizia Rovere-Querini
ABSTRACT
Muscle regeneration is a hallmark of the idiopathic inflammatory myopathies (IIMs), a group of autoimmune disorders that are characterized by leukocyte infiltration and dysfunction of the skeletal muscle. Despite detailed studies describing the clinical and histopathologic features of IIMs, the immunopathogenesis of these disorders remains undefined. The aim of this study was to investigate the immunopathologic processes of autoimmune myositis in experimental murine models. Expression of the autoantigen histidyl-transfer RNA synthetase (HisRS) was analyzed in mice with acutely injured or dystrophic muscles, in inflammatory leukocytes, and in purified satellite cells. Anti-HisRS antibodies and myositis induction were assessed in mice after muscle injury and immunization with apoptotic satellite cells or C2C12 myoblasts, in the presence or absence of the Toll-like receptor 7 (TLR-7) agonist R848. Muscle necrosis, leukocyte infiltration, and myofiber regeneration induced by toxic agents (cardiotoxin or glycerol) or promoted by genetic disruption of the α-sarcoglycan/dystrophin complex in mice were uniformly associated with up-regulated expression of HisRS. Although regenerating myofibers and purified satellite cells are known to show increased expression of HisRS in these settings, anti-HisRS antibodies were not detectable. However, intramuscular immunization with ultraviolet B-irradiated, HisRS-expressing apoptotic myoblasts in the presence of R848 triggered the production of anti-HisRS IgG antibodies as well as persistent lymphocyte infiltration and prolonged/delayed muscle regeneration. Conversely, intramuscular administration of R848 alone or in combination with living or postapoptotic/necrotic myoblasts failed to generate this myositis phenotype. In the presence of TLR/adjuvant signals and underlying muscle injury, apoptotic myogenic precursors expressing high levels of autoantigen can provoke autoantibody formation and lymphocytic infiltration of muscle tissue, effectively replicating the features of IIM.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Resiquimod, ≥98% (HPLC)
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%