Skip to Content
Merck
All Photos(1)

Key Documents

16911

Supelco

Hydrogen peroxide solution

30 % (w/w), for ultratrace analysis

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€126.00
1 L
€499.00
5 L
€1,070.00

€126.00


Estimated to ship on02 April 2025


Request a Bulk Order

Select a Size

Change View
250 ML
€126.00
1 L
€499.00
5 L
€1,070.00

About This Item

Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01
Beilstein:
3587191
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.24

€126.00


Estimated to ship on02 April 2025


Request a Bulk Order

grade

for inorganic trace analysis
for ultratrace analysis

Quality Level

vapor pressure

23.3 mmHg ( 30 °C)

form

liquid

shelf life

~5 yr

concentration

25-35%
30 % (w/w)

density

1.11 g/mL at 20 °C

anion traces

bromide (Br-): ≤10 μg/kg
chloride (Cl-): ≤25 μg/kg
fluoride (F-): ≤1000 μg/kg
nitrate (NO3-): ≤250 μg/kg
phosphate (PO43-): ≤25 μg/kg
sulfate (SO42-): ≤50 μg/kg

cation traces

Ag: ≤0.01 μg/kg
Al: ≤0.2 μg/kg
As: ≤0.05 μg/kg
Au: ≤0.01 μg/kg
B: ≤0.3 μg/kg
Ba: ≤0.05 μg/kg
Be: ≤0.01 μg/kg
Bi: ≤0.01 μg/kg
Ca: ≤2 μg/kg
Cd: ≤0.01 μg/kg
Ce: ≤0.01 μg/kg
Co: ≤0.01 μg/kg
Cr: ≤0.05 μg/kg
Cs: ≤0.01 μg/kg
Cu: ≤0.05 μg/kg
Dy: ≤0.01 μg/kg
Er: ≤0.01 μg/kg
Eu: ≤0.01 μg/kg
Fe: ≤0.2 μg/kg
Ga: ≤0.01 μg/kg
Gd: ≤0.01 μg/kg
Ge: ≤0.01 μg/kg
Hf: ≤0.01 μg/kg
Hg: ≤0.1 μg/kg
Ho: ≤0.01 μg/kg
In: ≤0.01 μg/kg
Ir: ≤0.01 μg/kg
K: ≤0.5 μg/kg
La: ≤0.01 μg/kg
Li: ≤0.01 μg/kg
Lu: ≤0.01 μg/kg
Mg: ≤0.2 μg/kg
Mn: ≤0.01 μg/kg
Mo: ≤0.01 μg/kg
N: ≤2000 μg/kg
Na: ≤0.5 μg/kg
Nb: ≤0.01 μg/kg
Nd: ≤0.01 μg/kg
Ni: ≤0.05 μg/kg
Os: ≤0.05 μg/kg
Pb: ≤0.1 μg/kg
Pd: ≤0.01 μg/kg
Pr: ≤0.01 μg/kg
Pt: ≤0.1 μg/kg
Rb: ≤0.01 μg/kg
Re: ≤0.01 μg/kg
Rh: ≤0.01 μg/kg
Ru: ≤0.01 μg/kg
Sb: ≤0.02 μg/kg
Sc: ≤0.05 μg/kg
Se: ≤0.05 μg/kg
Sm: ≤0.01 μg/kg
Sn: ≤0.01 μg/kg
Sr: ≤0.01 μg/kg
Tb: ≤0.01 μg/kg
Te: ≤0.01 μg/kg
Th: ≤0.01 μg/kg
Ti: ≤0.1 μg/kg
Tl: ≤0.01 μg/kg
Tm: ≤0.01 μg/kg
U: ≤0.01 μg/kg
V: ≤0.01 μg/kg
W: ≤0.02 μg/kg
Y: ≤0.01 μg/kg
Yb: ≤0.01 μg/kg
Zn: ≤0.2 μg/kg
Zr: ≤0.01 μg/kg

SMILES string

OO

storage temp.

2-8°C

InChI

1S/H2O2/c1-2/h1-2H

InChI key

MHAJPDPJQMAIIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Smartphone-Integrated Automated Sensor Employing Electrochemically Engineered 3D Bimetallic Nanoflowers for Hydrogen Peroxide Quantification in Milk: This innovative study demonstrates the application of high-purity hydrogen peroxide in the development of a sensor for detecting hydrogen peroxide levels in milk, integrating advanced technology for food safety and quality assurance (Kumari et al., 2024).
  • Assessment of new hydrogen peroxide activators in water and comparison of their active species toward contaminants of emerging concern: This study uses hydrogen peroxide solutions to evaluate new activators for water treatment processes, demonstrating their critical role in environmental testing and remediation efforts (Farinelli et al., 2024).

  • Albumin suppresses oxidation of Ti-Nb alloy in the simulated inflammatory environment: In material science research, hydrogen peroxide solutions are applied to study the oxidative resistance of biomedical alloys, highlighting their importance in developing durable and safe medical implants (Sotniczuk et al., 2024).



Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Shoichi Iriguchi et al.
Blood, 125(2), 370-382 (2014-10-29)
Although overexpression of T-bet, a master transcription factor in type-1 helper T lymphocytes, has been reported in several hematologic and immune diseases, its role in their pathogenesis is not fully understood. In the present study, we used transgenic model mice
Osamu Sakai et al.
Investigative ophthalmology & visual science, 56(1), 538-543 (2015-01-13)
The purpose of the present study was to investigate the role of glutathione peroxidase 4 (GPx4) in conjunctival epithelial cells. An immortalized human conjunctival epithelial cell line was used. Cells were transfected with catalase, GPx1, GPx4, SOD1, SOD2, or control
Arpád Dobolyi et al.
Brain structure & function, 220(1), 135-151 (2013-10-03)
SUCLA2 encodes the ATP-forming β subunit (A-SUCL-β) of succinyl-CoA ligase, an enzyme of the citric acid cycle. Mutations in SUCLA2 lead to a mitochondrial disorder manifesting as encephalomyopathy with dystonia, deafness and lesions in the basal ganglia. Despite the distinct
Martin Augsten et al.
Cancer research, 74(11), 2999-3010 (2014-04-09)
Cancer-associated fibroblasts (CAF) stimulate tumor growth and metastasis. Signals supporting CAF function are thus emerging as candidate therapeutic targets in the tumor microenvironment. The chemokine CXCL14 is a potent inducer of CAF protumorigenic functions. This study is aimed at learning
Koen van der Maaden et al.
Pharmaceutical research, 31(7), 1846-1854 (2014-01-29)
The aim of the study was to develop a cheap and fast method to produce hollow microneedles and an applicator for injecting vaccines into the skin at a pre-defined depth and test the applicability of the system for dermal polio

Questions

1–7 of 7 Questions  
  1. Is this product stabilized/inhibited like 216713?

    1 answer
    1. This product is not indicated as containing a stabilizer.

      Helpful?

  2. Is this solution, product 16911, ISO certified?

    1 answer
    1. This product has been assigned a quality level of 100 and is not considered to be ISO-certified. Please see the link below to review further information regarding assigned product quality levels:
      https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management/m-clarity-program

      Helpful?

  3. What is the pH of a 30% hydrogen peroxide solution?

    1 answer
    1. The pH of a 30 % hydrogen peroxide solution is typically between 3 and 4.

      Helpful?

  4. What is the molarity of the 30% hydrogen peroxide solution?

    1 answer
    1. The molarity of a 30 % hydrogen peroxide solution is 9.8 M based on a density of 1.11 g/ml.

      Helpful?

  5. How stable is the 30 % hydrogen peroxide solution?

    1 answer
    1. Although dilute solutions of hydrogen peroxide have a limited shelf life, the 30% solution (when properly stored) has a shelf life measured in years. Please see the certificate of analysis for the recommend retest date for a particular lot of Product No. 16911.

      Helpful?

  6. Can hydrogen peroxide be detected spectrophotometrically?

    1 answer
    1. Hydrogen peroxide has a molar extinction coefficient of 43.6 at 240 nm.

      Helpful?

  7. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service