Merck
  • OH-initiated photooxidations of 1-pentene and 2-methyl-2-propen-1-ol: mechanism and yields of the primary carbonyl products.
OH-initiated photooxidations of 1-pentene and 2-methyl-2-propen-1-ol: mechanism and yields of the primary carbonyl products.
Chemphyschem : a European journal of chemical physics and physical chemistry (2014-09-05)
Silvina A Peirone, Pablo M Cometto, Silvia I Lane
ABSTRACT
The products of the gas-phase reactions of OH radicals with 1-pentene and 2-methyl-2-propen-1-ol (221MPO) at T=298±2 K and atmospheric pressure were investigated by using a 4500 L atmospheric simulation chamber that was built especially for this work. The molar yield of butyraldehyde was 0.74±0.12 mol for the reaction of 1-pentene. This work provides the first product molar yield determination of formaldehyde (0.82±0.12 mol), 1-hydroxypropan-2-one (0.84±0.13 mol), and methacrolein (0.078±0.012 mol) from the reaction of 221MPO with OH radicals. The mechanism of this reaction is discussed in relation to the experimental results. Additionally, taking into consideration the complex mechanism, the rate coefficients of the reactions of OH with formaldehyde, 1-hydroxypropan-2-one, and methacrolein were derived at atmospheric pressure and T=298±2 K.; the obtained values were (8.9±1.6)×10(-12) , (2.4±1.4)×10(-12) , and (22.9±2.3)×10(-12) cm(3) molecule(-1) s(-1) , respectively.
Product Description
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga, metal alloy fiber
Supelco
SPME StableFlex fiber assortment kit, for use with manual holder
Supelco
SPME fiber assortment kit 2, for use with autosampler, size 24 ga
Supelco
SPME fiber assortment kit 4, for use with manual holder, size 24 ga
Supelco
SPME fiber assortment kit 2, for use with autosampler, needle size 23 ga
Supelco
SPME StableFlex fiber assortment kit, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assortment kit 4, for use with autosampler, needle size 23 ga
Supelco
SPME StableFlex fiber assortment kit, needle size 23 ga, StableFlex fiber, for use with autosampler
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assortment kit 2, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assortment kit 4, for use with autosampler, size 24 ga
Supelco
SPME fiber assortment kit 1, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 24 ga
Sigma-Aldrich
1-Pentene, 98%
Sigma-Aldrich
7-Amino-1,3-naphthalenedisulfonic acid monopotassium salt monohydrate, BioReagent, suitable for fluorescence, ≥98.0% (T)
Supelco
1-Pentene, analytical standard
Sigma-Aldrich
1-Pentene, ≥98.5% (GC)
Supelco
SPME fiber assortment kit 5, needle size 23 ga, for use with autosampler
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Supelco
Butyraldehyde, analytical standard, contains ~0.1% 2,6-di-tert-butyl-4-methylphenol and ~1% water as stabilizer
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, for use with autosampler, needle size 23 ga, metal alloy fiber
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (1cm), needle size 23 ga, Autosampler, pk of 1 ×, plain gray hub
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 24 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), fused silica fiber, df 65 μm(PDMS/DVB, for use with manual holder, needle size 23 ga
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 24 ga
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub