Merck
  • Versatile ene-thiol photoclick reaction for preparation of multimodal monolithic silica capillary columns.
Versatile ene-thiol photoclick reaction for preparation of multimodal monolithic silica capillary columns.
Journal of chromatography. A (2014-09-25)
A Marechal, A Laaniste, R El-Debs, V Dugas, C Demesmay
ABSTRACT
This paper presents a photografting process of monolithic silica capillary columns based on the ene-thiol click chemistry. This study is performed on a "generic" vinyl-functionalized silica monolith (Hmin 6±1μm). The photoclick reaction is investigated using different thiol monomers (octadecanethiol, cysteine and sodium mercaptoethanesulfonate) to prepare capillary columns dedicated to various chromatographic modes (reversed-phase, HILIC and strong cation exchange). Whatever the monomer used, the photografting reaction is achieved in less than 5min with a relatively high thiol monomer content. This allows preparing highly retentive and efficient monolithic columns while avoiding polymerization and/or column clogging. In addition to the aforementioned properties (duration, versatility, efficiency), this photo-triggered chemical reaction allows addressing several appropriate surface functionalizations inside a single monolithic column in order to prepare nanovolume multimodal capillary columns. A multimodal biphasic monolithic column with a 1cm length cation-exchange segment followed by a 9cm length reversed-phase segment (SCX-RP) is prepared through two successive photografting reactions using a UV-mask to localize the reactions. This multimodal biphasic column is investigated using a model sample for the selective fractionation and separation of cationic and neutral compounds and is applied to the on-line preconcentration and separation of β-blockers.
Product Description
Gemcitabine impurity A, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Fluorouracil impurity C, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Benzophenone, purified by sublimation, ≥99%
Supelco
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Toluene, analytical standard
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Melting point standard 79-81°C, analytical standard
Sigma-Aldrich
Vinyltrimethoxysilane, 98%
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Sigma-Aldrich
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Tetramethyl orthosilicate, 98%
Sigma-Aldrich
Vinyltrimethoxysilane, 97%
Supelco
Benzene, analytical standard
Sigma-Aldrich
Tetramethyl orthosilicate, ≥99%
Sigma-Aldrich
Benzophenone, ReagentPlus®, 99%
Sigma-Aldrich
Tetramethyl orthosilicate, purum, ≥98.0% (GC)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Adenosine
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%