Skip to Content
Merck
All Photos(1)

Key Documents

27602

Sigma-Aldrich

Caffeine

anhydrous, tested according to Ph. Eur.

Synonym(s):

Coffeinum, 1,3,7-Trimethylxanthine

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€107.00

€107.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
€107.00

About This Item

Empirical Formula (Hill Notation):
C8H10N4O2
CAS Number:
Molecular Weight:
194.19
Beilstein:
17705
EC Number:
MDL number:
UNSPSC Code:
12352210
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous

€107.00


Check Cart for Availability

Request a Bulk Order

grade

anhydrous

Quality Level

Agency

EPA 1694
USP/NF
tested according to Ph. Eur.

form

powder

mp

234-236.5 °C (lit.)

solubility

H2O: soluble 18.7 g/L at 16 °C

application(s)

environmental

SMILES string

CN1C(=O)N(C)c2ncn(C)c2C1=O

InChI

1S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3

InChI key

RYYVLZVUVIJVGH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Caffeine has been used as a radiosensitizing agent utilized for studying its impact in inhibiting the catalytic activity of ATM and ATR Kinase.[1] CTGF is an important modulator protein of fibrogenic TGF-β. Hence, this product has also been used for examining the caffeine-dependent regulation of CTGF expression in rat hepatocytes.[2]

Biochem/physiol Actions

Caffeine is a central nervous system stimulant, adenosine receptor antagonist as well as adenosine 3′,5′-cyclic monophosphate (cAMP) phosphodiesterase inhibitor. It stimulates the sarcoplasmic reticulum (SR) Ca2+-release channel which facilitates the excitation-contraction coupling in cardiac muscle.[3] It has also been reported to affect cellular calcium levels, releasing calcium from intracellular stores. Caffeine overrides the cell cycle effects of various chemicals such as protease inhibitors, preventing apoptosis; and it has been shown to inhibit cellular DNA repair mechanisms.[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J N Sarkaria et al.
Cancer research, 59(17), 4375-4382 (1999-09-15)
Caffeine exposure sensitizes tumor cells to ionizing radiation and other genotoxic agents. The radiosensitizing effects of caffeine are associated with the disruption of multiple DNA damage-responsive cell cycle checkpoints. The similarity of these checkpoint defects to those seen in ataxia-telangiectasia
E Rousseau et al.
The American journal of physiology, 256(2 Pt 2), H328-H333 (1989-02-01)
Caffeine is thought to affect excitation-contraction coupling in cardiac muscle by activating the sarcoplasmic reticulum (SR) Ca2+-release channel. The effect of caffeine at the single channel level was studied by incorporating canine cardiac SR vesicles into planar lipid bilayers. Cardiac
Lukas Kovar et al.
Pharmaceuticals (Basel, Switzerland), 15(2) (2022-02-27)
Static in vitro permeation experiments are commonly used to gain insights into the permeation properties of drug substances but exhibit limitations due to missing physiologic cell stimuli. Thus, fluidic systems integrating stimuli, such as physicochemical fluxes, have been developed. However
C P Selby et al.
Proceedings of the National Academy of Sciences of the United States of America, 87(9), 3522-3525 (1990-05-01)
Caffeine potentiates the mutagenic and lethal effects of genotoxic agents. It is thought that this is due, at least in some organisms, to inhibition of DNA repair. However, direct evidence for inhibition of repair enzymes has been lacking. Using purified
Jueng-Eun Im et al.
Pharmaceutics, 13(5) (2021-05-06)
Dermal absorption of chemicals is a key factor in risk assessment. This study investigated the effects of different amounts of application on dermal absorption and suggested an appropriate application dose for proper dermal absorption. Caffeine and testosterone were chosen as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service