Merck
  • Alpha amylase enzyme inhibitory and anti-inflammatory effect of Lawsonia inermis.
Alpha amylase enzyme inhibitory and anti-inflammatory effect of Lawsonia inermis.
Pakistan journal of biological sciences : PJBS (2014-02-11)
Hasan Imam, Nasir Uddin Mahbub, Md Forhad Khan, Humayera Kabir Hana, Md Moklesur Rahman Sarker
ABSTRACT
Previously it was reported elsewhere that Lawsonia inermis have anti-inflammatory and analgesic effect in experimental animals. The in vitro porcine alpha amylase inhibitory effect was investigated of this plant methanolic extracts and consequently hypoglycemic effect by quantitatively determining the maltose from the maltose standard curve while the anti-inflammatory effect by acetic acid induced writhing test in mice. Acarbose (10 microg mL(-1)) and Diclofenac sodium (20 mg kg(-1)) were used as reference hypoglycemic and anti-inflammatory drugs, respectively, for this study. The methanolic leaves extract of the plant significantly inhibited (60.97% compared to untreated) enzymatic activity of the amylase at 10 microg mL(-1) dose (p < 0.05) also reduced the chemically induced nociceptive pain stimuli significantly at all doses (p < 0.01). Carbohydrates, glycosides, flavonoids, saponins and tannins were found to have in phytochemical screening of the extract which are thought to bring these effects. For the conclusive purpose, it is suggesting from the result that the pharmacological properties of this Lawsonia inermis can elicit hypoglycemic effect by inhibiting alpha-amylase enzyme and can reduce neurogenic pain stimulus. It gives the notion that how this group of patient would be therapeutically benefitted by decreasing both these effects by the same agent which is easy available.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, ≥400 units/mg protein (Lowry)
Supelco
Methanol, analytical standard
Sigma-Aldrich
α-Amylase, heat-stable, solution, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from porcine pancreas, PMSF Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from porcine pancreas, Type I-A, PMSF treated, saline suspension, 700-1400 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
Sigma-Aldrich
Taka-Diastase from Aspergillus oryzae, powder, slightly beige, ~100 U/mg
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~50 U/mg
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
α-Amylase from Bacillus sp., liquid
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~380 U/mg
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%