Merck
  • Palladium-catalyzed cyclization of o-alkynyltrifluoroacetanilides followed by isocyanide insertion: synthesis of 2-substituted 1H-indole-3-carboxamides.
Palladium-catalyzed cyclization of o-alkynyltrifluoroacetanilides followed by isocyanide insertion: synthesis of 2-substituted 1H-indole-3-carboxamides.
Chemical communications (Cambridge, England) (2012-06-21)
Ziwei Hu, Dongdong Liang, Jiaji Zhao, Jinbo Huang, Qiang Zhu
ABSTRACT
A base-controlled synthesis of 2-substituted secondary and tertiary 1H-indole-3-carboxamides through PdCl(2)-catalyzed cyclization of o-alkynyltrifluoroacetanilides followed by isocyanide insertion has been developed. The reaction proceeds smoothly at ambient temperature using O(2) in air as the sole oxidant of the palladium catalyst.
Sigma-Aldrich
Palladium(II) chloride solution, 5 wt. % in 10 wt. % HCl
Sigma-Aldrich
Palladium(II) chloride ChemBeads
Sigma-Aldrich
Palladium(II) chloride, anhydrous, 60% Pd basis
Sigma-Aldrich
Palladium(II) chloride, ≥99.9%
Sigma-Aldrich
Palladium(II) chloride, anhydrous, 59-60% palladium (Pd) basis
Sigma-Aldrich
Palladium(II) chloride, 99.995%
Sigma-Aldrich
Palladium(II) chloride, ReagentPlus®, 99%