Merck
  • Synthesis, kinetic studies and pharmacological evaluation of mutual azo prodrugs of 5-aminosalicylic acid for colon-specific drug delivery in inflammatory bowel disease.
Synthesis, kinetic studies and pharmacological evaluation of mutual azo prodrugs of 5-aminosalicylic acid for colon-specific drug delivery in inflammatory bowel disease.
European journal of medicinal chemistry (2009-05-16)
Suneela S Dhaneshwar, Neha Gairola, Mini Kandpal, Gaurav Vadnerkar, Lokesh Bhatt, Badal Rathi, S S Kadam
ABSTRACT
Colon-specific mutual azo prodrugs of 5-aminosalicylic acid with essential amino acids were synthesized for the management of inflammatory bowel disease. The structures were confirmed by elemental and spectral analyses. 85-88% release of 5-aminosalicylic acid was achieved in rat fecal matter with half-lives ranging from 140 to 160 min, following first order kinetics. The prodrugs exhibited comparable ameliorating effect as that of sulfasalazine on trinitrobenzenesulfonic acid-induced experimental colitis in rats with a better safety profile.
Product Description
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
SAFC
L-Tyrosine
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
D-Phenylalanine, ≥98% (HPLC)
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
5-Aminosalicylic acid, 95%
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
5-Aminosalicylic acid, ≥99%
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)