Merck
  • Abundance of d-2-hydroxyglutarate in G2/M is determined by FOXM1 in mutant IDH1-expressing cells.
Abundance of d-2-hydroxyglutarate in G2/M is determined by FOXM1 in mutant IDH1-expressing cells.
FEBS letters (2019-06-19)
Kancharana Bala Bhaskara Rao, Kumar Katragunta, Uma Maheswara Sarma, Nishant Jain
ABSTRACT
Isocitrate dehydrogenases (IDHs) are metabolic enzymes that are mutated in several cancers, resulting in overproduction of d-2-hydroxyglutarate (D-2HG). However, the signalling pathways and factors that regulate mutant IDHs or their metabolites remain elusive. Here, we report that in synchronized cells and cells treated with anti-mitotic agents, wild-type and mutant IDH proteins are induced maximally in G2/M. Moreover, mutant IDH1-expressing cells arrested in G2/M harbour high D2HG levels. Genetic or pharmacological perturbation of Forkhead box protein M1 (FOXM1) abrogates the levels of IDH1 mRNA, protein and D2HG in G2/M. Conversely, overexpression of FOXM1 or hyperactive FOXM1 activates the IDH1 promoter and increases the abundance of its protein levels. In summary, our results show that in G2/M, higher D2HG levels are dependent on FOXM1-mediated transcription of IDH1.
Sigma-Aldrich
Anti-Cyclin B1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Polyethylenimine, branched, average Mw ~25,000 by LS, average Mn ~10,000 by GPC, branched
Sigma-Aldrich
D-α-Hydroxyglutaric acid disodium salt, ≥95% (GC)
Sigma-Aldrich
Podophyllotoxin
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-GFP antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
Thiostrepton from Streptomyces azureus, ≥90% (HPLC)
Sigma-Aldrich
Anti-HA−Peroxidase antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Cyclin A antibody produced in mouse, clone CY-A1, ascites fluid
Sigma-Aldrich
Combretastatin A4, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
AGI-6780, ≥98% (HPLC)
Sigma-Aldrich
L-α-Hydroxyglutaric acid disodium salt, ≥98.0% (GC)
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
L-Glutamine–Penicillin–Streptomycin solution, L-glutamine 200 mM, streptomycin 10 mg/mL, penicillin 10,000 units, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Monoclonal Anti-T7 tag antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)