Skip to Content
Merck

Skip To

C4710

Monoclonal Anti-Cyclin A antibody produced in mouse

clone CY-A1, ascites fluid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
0.2 mL
Check Cart for Availability
€ 583,00
€ 495,55

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
CY-A1, monoclonal
Application:
ARR, ICC, WB
Citations:
48

€ 495,55

List Price€ 583,00Save 15%
Web-Only Promotion

Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1590

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

CY-A1, monoclonal

mol wt

antigen 60 kDa

contains

15 mM sodium azide

species reactivity

mouse, human, monkey, bovine

technique(s)

immunocytochemistry: suitable, microarray: suitable, western blot: 1:1,000 using a mouse NIH 3T3 cell extract

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCNA2(890)
mouse ... Ccna2(12428)

General description

Cyclin A is a nuclear protein during S phase and it binds both cdk2 and cdc2. The availability of monoclonal antibody reacting specifically with cyclin A enables the subcellular detection and localization of cyclin A and the measurement of relative differences in cyclin A levels as a function of cell cycle phase.

Immunogen

recombinant bovine cyclin A.

Application

Monoclonal Anti-Cyclin A antibody produced in mouse is suitable for:
  • immunocytochemistry
  • microarray
  • western blotting at a dilution of 1:1,000 using a mouse NIH 3T3 cell extract

Biochem/physiol Actions

The antibody recognizes an epitope present in amino acids 162-433 of bovine cyclin A. No cross-reactivity is detected with other cyclins (cyclin B, D, or E).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C7464C8218WH0008900M2
species reactivity

mouse, human, monkey, bovine

species reactivity

rat, mouse, human, monkey

species reactivity

rat, mouse, human

species reactivity

human

clone

CY-A1, monoclonal

clone

DCS-6, monoclonal

clone

DCS-31, monoclonal

clone

4A11-5B5, monoclonal

antibody form

ascites fluid

antibody form

ascites fluid

antibody form

ascites fluid

antibody form

purified immunoglobulin

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Gene Information

human ... CCNA2(890)
mouse ... Ccna2(12428)

Gene Information

human ... CCND1(595)
mouse ... Ccnd1(12443)
rat ... Ccnd1(58919)

Gene Information

human ... CDK4(1019)
mouse ... Cdk4(12567)
rat ... Cdk4(94201)

Gene Information

human ... CCNA1(8900)

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters