Skip to Content
Merck

Skip To

SML0895

AGI-6780

≥98% (HPLC)

Synonym(s):

N-Cyclopropyl-4-(3-thienyl)-3-[[[[3-(trifluoromethyl)phenyl]amino]carbonyl]amino]-benzenesulfonamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 145,00
€ 123,25
25 mg
Check Cart for Availability
€ 420,00

About This Item

Empirical Formula (Hill Notation):
C21H18F3N3O3S2
CAS Number:
Molecular Weight:
481.51
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

€ 123,25

List Price€ 145,00Save 15%
Web-Only Promotion

Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc(ccc1)NC(=O)Nc2c(ccc(c2)[S](=O)(=O)NC4CC4)c3c[s]cc3

InChI

1S/C21H18F3N3O3S2/c22-21(23,24)14-2-1-3-16(10-14)25-20(28)26-19-11-17(32(29,30)27-15-4-5-15)6-7-18(19)13-8-9-31-12-13/h1-3,6-12,15,27H,4-5H2,(H2,25,26,28)

InChI key

CCAWRGNYALGPQH-UHFFFAOYSA-N

Biochem/physiol Actions

AGI-6780 is a selective inhibitor of IDH2/R140Q a tumor-associated mutant of isocitrate dehydrogenase IDH2.
AGI-6780 is a selective inhibitor of IDH2/R140Q a tumor-associated mutant of isocitrate dehydrogenase IDH2. Several human cancers have somatic point mutations in the genes encoding isocitrate dehydrogenases 1 and 2 (IDH1 and IDH2), confering a gain-of-function in cancer cells in which the mutant enzymes produce the oncometabolite (R)-2-hydroxyglutarate, [(R)-2HG]. (R)-2HG inhibits histone- and DNA-modifying enzymes, resulting in altered gene expression, histone and DNA hypermethylation, and inducing a block in cellular differentiation that promotes tumorigenesis. AGI-6780 reduced (R)-2HG levels in cell lines overexpressing IDH2/R140Q with an EC50 of 20 nM, and induced differentiation of TF-1 erythroleukemia and primary human acute myelogenous leukemia cells in vitro. AGI-6780 binds allosterically at the dimer interface of IDH2. AGI-6780 shows good selectivity against other dehydrogenases, including the closely related IDH1-WT or R132H mutant enzymes.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1990SML2657SML1738
form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

-

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

room temp

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 25 mg/mL, clear

color

white to light brown

color

white to beige

color

white to beige

color

white to beige


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters