Skip to Content
Merck
All Photos(1)

Key Documents

V900535

Sigma-Aldrich

1-(2-Hydroxyethyl)piperazine

Vetec, reagent grade, 98%

Synonym(s):

2-Piperazinoethanol, Piperazine-1-ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N2O
CAS Number:
Molecular Weight:
130.19
Beilstein:
104361
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

refractive index

n20/D 1.506 (lit.)

bp

246 °C (lit.)

density

1.061 g/mL at 25 °C (lit.)

SMILES string

OCCN1CCNCC1

InChI

1S/C6H14N2O/c9-6-5-8-3-1-7-2-4-8/h7,9H,1-6H2

InChI key

WFCSWCVEJLETKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana I Loureiro et al.
Xenobiotica; the fate of foreign compounds in biological systems, 45(9), 828-839 (2015-04-29)
1. This study explores the impact of permeability and P-glycoprotein (P-gp) efflux, upon brain exposure to etamicastat, a new dopamine-β-hydroxylase (DBH) inhibitor and consequently brain levels of catecholamines. 2. Brain exposure to etamicastat (10 mg/kg), following intravenous administration to mice, was
Katalin F Medzihradszky et al.
Analytical chemistry, 87(5), 3064-3071 (2015-02-06)
Beam-type collision-induced dissociation (CID) data of intact glycopeptides isolated from mouse liver tissue are presented to illustrate characteristic fragmentation of differentially sialylated glycopeptides. Eight glycoforms of an O-linked glycopeptide from Nucleobindin-1 are distinguished on the basis of the precursor masses
Patrice N Mimche et al.
Hepatology (Baltimore, Md.), 62(3), 900-914 (2015-03-19)
Beyond the well-defined role of the Eph (erythropoietin-producing hepatocellular) receptor tyrosine kinases in developmental processes, cell motility, cell trafficking/adhesion, and cancer, nothing is known about their involvement in liver pathologies. During blood-stage rodent malaria infection we have found that EphB2
Yang Zhang et al.
PloS one, 10(6), e0128555-e0128555 (2015-06-05)
It has been well established that the architecture of chromatin in cell nuclei is not random but functionally correlated. Chromatin damage caused by ionizing radiation raises complex repair machineries. This is accompanied by local chromatin rearrangements and structural changes which
William H Parker et al.
Biochemical and biophysical research communications, 458(2), 262-267 (2015-02-04)
Intracellular vitamin C, or ascorbic acid, has been shown to prevent the apoptosis of cultured vascular pericytes under simulated diabetic conditions. We sought to determine the mechanism by which ascorbate is transported into pericytes prior to exerting this protective effect.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service