Merck
The receptor tyrosine kinase EphB2 promotes hepatic fibrosis in mice.
Hepatology (Baltimore, Md.) (2015-03-19)
Patrice N Mimche, Lauren M Brady, Christian F Bray, Choon M Lee, Manoj Thapa, Thayer P King, Kendra Quicke, Courtney D McDermott, Sylvie M Mimche, Arash Grakoui, Edward T Morgan, Tracey J Lamb
ABSTRACT
Beyond the well-defined role of the Eph (erythropoietin-producing hepatocellular) receptor tyrosine kinases in developmental processes, cell motility, cell trafficking/adhesion, and cancer, nothing is known about their involvement in liver pathologies. During blood-stage rodent malaria infection we have found that EphB2 transcripts and proteins were up-regulated in the liver, a result likely driven by elevated surface expression on immune cells including macrophages. This was significant for malaria pathogenesis because EphB2(-/-) mice were protected from malaria-induced liver fibrosis despite having a similar liver parasite burden compared with littermate control mice. This protection was correlated with a defect in the inflammatory potential of hepatocytes from EphB2(-/-) mice resulting in a reduction in adhesion molecules, chemokine/chemokine receptor RNA levels, and infiltration of leukocytes including macrophages/Kupffer cells, which mediate liver fibrosis during rodent malaria infections. These observations are recapitulated in the well-established carbon tetrachloride model of liver fibrosis in which EphB2(-/-) carbon tetrachloride-treated mice showed a significant reduction of liver fibrosis compared to carbon tetrachloride-treated littermate mice. Depletion of macrophages by clodronate-liposomes abrogates liver EphB2 messenger RNA and protein up-regulation and fibrosis in malaria-infected mice. During rodent malaria, EphB2 expression promotes malaria-associated liver fibrosis; to our knowledge, our data are the first to implicate the EphB family of receptor tyrosine kinases in liver fibrosis or in the pathogenesis of malaria infection.
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Carbon tetrachloride, anhydrous, ≥99.5%
Sigma-Aldrich
1-(2-Hydroxyethyl)piperazine, 98%
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
SAFC
L-Glutamine
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
Hematoxylin, certified by the BSC