Skip to Content
Merck
All Photos(2)

Key Documents

V900418

Sigma-Aldrich

Biotin

Vetec, reagent grade, ≥99%

Synonym(s):

D-Biotin, Bios II, Coenzyme R, Vitamin B7, Vitamin H

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O3S
CAS Number:
Molecular Weight:
244.31
Beilstein:
86838
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

≥99%

form

powder

color

white to off-white

mp

231-233 °C (lit.)

storage temp.

2-8°C

SMILES string

[H][C@]12CS[C@@H](CCCCC(O)=O)[C@@]1([H])NC(=O)N2

InChI

1S/C10H16N2O3S/c13-8(14)4-2-1-3-7-9-6(5-16-7)11-10(15)12-9/h6-7,9H,1-5H2,(H,13,14)(H2,11,12,15)/t6-,7-,9-/m0/s1

InChI key

YBJHBAHKTGYVGT-ZKWXMUAHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Biotin (vitamin B7 or vitamin H) is a water-soluble B complex vitamin, which consists of an uredo ring fused by a tetrahydrothiophene ring.[1] It is available in various foods, like whole grains, nuts, legumes, unpolished rice and egg yolk. Biotin is also produced by the normal gut flora.[2]

Biochem/physiol Actions

Biotin is a vital cofactor for carboxylase enzymes in several metabolic pathways. It also functions as a coenzyme in the metabolism of fatty acids, isoleucine and valine. Biotin assists the transfer of carbon dioxide and also sustains a steady blood sugar level. Biotin is implicated in gluconeogenesis and citric acid cycle.[1] It is involved in keratin synthesis and hence is used as a supplement for skin, hair and nail growth.[2] Biotin is required for cell growth, production of fatty acids and the metabolism of fats and amino acids.[1]

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Vitamin H and their role in ruminant: A Review
Pour HA, et al.
Annals of Biological Research, 3(4), 1929-1933 (2012)
A Review of the Use of Biotin for Hair Loss
Patel DP, et al.
Skin Appendage Disorder, 3(3), 166-166 (2017)
Naruo Nikoh et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(28), 10257-10262 (2014-07-02)
Obligate insect-bacterium nutritional mutualism is among the most sophisticated forms of symbiosis, wherein the host and the symbiont are integrated into a coherent biological entity and unable to survive without the partnership. Originally, however, such obligate symbiotic bacteria must have
Tuan V Bui et al.
Neuron, 78(1), 191-204 (2013-04-16)
Accurate motor performance depends on the integration in spinal microcircuits of sensory feedback information. Hand grasp is a skilled motor behavior known to require cutaneous sensory feedback, but spinal microcircuits that process and relay this feedback to the motor system
Konstantinos Ampatzis et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(26), 10875-10886 (2013-06-28)
In vertebrates, spinal circuits drive rhythmic firing in motoneurons in the appropriate sequence to produce locomotor movements. These circuits become active early during development and mature gradually to acquire the flexibility necessary to accommodate the increased behavioral repertoire of adult

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service