Skip to Content
Merck
Biotin tested according to Ph. Eur.

Biotin

Biotin tested according to Ph. Eur.
Synonym(s):
D-Biotin, Bios II, Coenzyme R, Vitamin B7, Vitamin H, Biotin
Empirical Formula (Hill Notation):
C10H16N2O3S
CAS Number:
Molecular Weight:
244.31
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique