Skip to Content
Merck
All Photos(1)

Key Documents

V800394

Sigma-Aldrich

Sodium phosphate

AR, ≥95.5%

Synonym(s):

Sodium orthophosphate, Trisodium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na3PO4
CAS Number:
Molecular Weight:
163.94
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥95.5%
grade:
AR
form:
powder, crystals or granules

grade

AR

product line

Vetec

Assay

≥95.5%

form

powder, crystals or granules

pH

11 (20 °C)

cation traces

Fe: ≤0.001%
heavy metals (as Pb): ≤0.001%

SMILES string

[Na+].[Na+].[Na+].[O-]P([O-])([O-])=O

InChI

1S/3Na.H3O4P/c;;;1-5(2,3)4/h;;;(H3,1,2,3,4)/q3*+1;/p-3

InChI key

RYFMWSXOAZQYPI-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qin Wang et al.
International journal of epidemiology, 44(2), 623-637 (2015-06-08)
The causal role of circulating sex hormone-binding globulin (SHBG) for type 2 diabetes is controversial. Information on the relations between SHBG and new biomarkers of cardiometabolic risk is scarce. We applied quantitative nuclear magnetic resonance metabolomics in three Finnish population-based
L Ren et al.
Nanotechnology, 26(28), 284001-284001 (2015-06-27)
The prospect for spatial imaging with mass spectroscopy at the level of the cell requires new means of cell extraction to conserve molecular structure. To this aim, we demonstrate a new laser extraction process capable of extracting intact biological entities
Stephanie J Bissel et al.
Journal of neuropathology and experimental neurology, 74(8), 767-777 (2015-06-27)
Human parechovirus 3 (HPeV3) is a picornavirus associated with neurologic disease in neonates. Human parechovirus 3 infection of preterm and term infants is associated with seizures and destructive periventricular white matter lesions. Despite unremarkable cerebrospinal fluid (CSF), HPeV3 RNA can
Montse Visa et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(7), 2970-2979 (2015-03-27)
The toxic effects of human islet amyloid polypeptide (IAPP) on pancreatic islets have been widely studied. However, much less attention has been paid to the physiologic actions of IAPP on pancreatic β cells, which secrete this peptide together with insulin
David H Kwan et al.
Journal of the American Chemical Society, 137(17), 5695-5705 (2015-04-15)
Blood transfusions are critically important in many medical procedures, but the presence of antigens on red blood cells (RBCs, erythrocytes) means that careful blood-typing must be carried out prior to transfusion to avoid adverse and sometimes fatal reactions following transfusion.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service