Skip to Content
Merck
All Photos(2)

Key Documents

935670

Sigma-Aldrich

Sodium phosphate tribasic

greener alternative

≥99.9% trace metals basis

Synonym(s):

Sodium orthophosphate, Trisodium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na3PO4
CAS Number:
Molecular Weight:
163.94
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

Assay

≥99.9% trace metals basis

form

(powder, crystals, or granules)

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤1500 (trace metals analysis)

color

white to off-white

mp

1583 °C

solubility

water: slightly soluble

density

2.54 g/cm3

greener alternative category

SMILES string

[Na+].[Na+].[Na+].[O-]P([O-])([O-])=O

InChI

1S/3Na.H3O4P/c;;;1-5(2,3)4/h;;;(H3,1,2,3,4)/q3*+1;/p-3

InChI key

RYFMWSXOAZQYPI-UHFFFAOYSA-K

General description

We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives. This product has been enhanced for energy efficency. Click here for more information.

Application

Sodium phosphate tribasic is used to synthesis a reproducible Ag3PO4/TiO2 visible light photocatalyst to improve the photocatalytic activity and stability of Ag3PO4[1]. It has also been employed as a mineralizer in the hydrothermal synthesis of uniform single-crystalline CeO2 nano-octahedrons and nanorods. It does not leave any impurities in the reaction system and makes it very easy to obtain and separate the octahedral and rod-like morphology[2].In addition, anodizing of Magnesium is studied in a solution containing sodium phosphate tribasic and KOH-aluminate to form anodic films of MgO and MgAl2O4 [3]. Microarc oxidation coatings on magnesium alloy were created in sodium phosphate-KOH electrolytes with NaAlO2. The addition of NaAlO2 to the base electrolyte increased the amount of spinel MgAl2O4 in the oxide coating[4].

In general, sodium phosphate tribasic is used as:

  1. component in the preparation of solid crystals of calcium hydroxyapatite substituted by strontium
  2. block buffer to study segmental and subcellular distribution of CFTR in the kidney
  3. pH control agent, water hardness precipitation, laboratory reagent, phosphatizing reagent.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of high efficiency and stable Ag3PO4/TiO2 visible light photocatalyst for the degradation of methylene blue and rhodamine B solutions
Yao W, et al.
Journal of Materials Chemistry, 22, 4050-4055 (2012)
Effects of NaAlO2 on structure and corrosion resistance of microarc oxidation coatings formed on AM60B magnesium alloy in phosphate-KOH electrolyte
Liang J, et al.
Surface and Coatings Technology, 199, 121-126 (2005)
Template-Free Hydrothermal Synthesis of CeO2Nano-octahedrons and Nanorods: Investigation of the Morphology Evolution
Lai Y, et al.
Crystal Growth & Design, 8, 1474?1477-1474?1477 (2008)
Anodizing of Pure Magnesium in KOH?Aluminate Solutions under Sparking
Khaselev O, et al.
Journal of the Electrochemical Society, 146, 1757-1757 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service