Skip to Content
Merck
All Photos(2)

Documents

V800380

Sigma-Aldrich

Sodium hexametaphosphate

LR, ≥67% P2O5 basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H6O18P6 · 6Na
CAS Number:
Molecular Weight:
617.82
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

LR

product line

Vetec

Assay

≥67% P2O5 basis

form

powder

anion traces

chloride (Cl-): ≤0.02%

cation traces

Fe: ≤0.03%
heavy metals (as Pb): ≤0.005%

SMILES string

[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[O-]P1(=O)OP([O-])(=O)OP([O-])(=O)OP([O-])(=O)OP([O-])(=O)OP([O-])(=O)O1

InChI

1S/6Na.H6O18P6/c;;;;;;1-19(2)13-20(3,4)15-22(7,8)17-24(11,12)18-23(9,10)16-21(5,6)14-19/h;;;;;;(H,1,2)(H,3,4)(H,5,6)(H,7,8)(H,9,10)(H,11,12)/q6*+1;/p-6

InChI key

GCLGEJMYGQKIIW-UHFFFAOYSA-H

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Megumu Sakoda et al.
Biomacromolecules, 19(8), 3280-3290 (2018-07-10)
We have developed a new hydrogel hemostat composed of hyaluronan (HA) conjugated with inorganic polyphosphate (PolyP). A hemostatic hydrogel, HAX-PolyP, was formed rapidly by mixing aldehyde-modified HA and hydrazide-modified HA conjugated with PolyP (HA-PolyP). Although the gelation rate decreased with
Yanli Tang et al.
Langmuir : the ACS journal of surfaces and colloids, 27(8), 4945-4955 (2011-03-17)
Three series of cationic oligo p-phenyleneethynylenes (OPEs) have been synthesized to study their structure-property relationships and gain insights into the transition from molecular to macromolecular properties. The absorbance maxima and molar extinction coefficients in all three sets increase with increasing
Deepak H Veeregowda et al.
European journal of oral sciences, 119(1), 21-26 (2011-01-20)
The visco-elasticity of salivary-protein films is related to mouthfeel, lubrication, biofilm formation, and protection against erosion and is influenced by the adsorption of toothpaste components. The thickness and the visco-elasticity of hydrated films (determined using a quartz crystal microbalance) of
Pauline Wanjugi et al.
Water research, 140, 200-210 (2018-05-02)
There is a growing interest for the use of coliphage as an alternative indicator to assess fecal pollution in recreational waters. Coliphage are a group of viruses that infect Escherichia coli and are considered as potential surrogates to infer the
Björn Drobot et al.
Nature communications, 9(1), 3643-3643 (2018-09-09)
Phase separation of mixtures of oppositely charged polymers provides a simple and direct route to compartmentalisation via complex coacervation, which may have been important for driving primitive reactions as part of the RNA world hypothesis. However, to date, RNA catalysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service