Skip to Content
Merck
All Photos(1)

Key Documents

V800352

Sigma-Aldrich

Silica gel

GF254, suitable for thin layer chromatography (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

product line

Vetec

form

solid

technique(s)

thin layer chromatography (TLC): suitable

impurities

10.0-15.0% calcium sulfate

bp

2230 °C

mp

>1600 °C

anion traces

chloride (Cl-): ≤0.02%

cation traces

Fe: ≤0.02%

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silica gel is an amorphous form of silicon dioxide (SiO2), which is widely used as a desiccant for moisture absorption, as a stationary phase in chromatographic applications, and also as a humidity indicator.

Application

Silica gel can be used as a stationary phase in thin-layer chromatography to separate a mixture of non-volatile compounds using a single solvent or a mixture of solvents as mobile phase.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Reversed-phase liquid chromatographic separation of enantiomers on polysaccharide type chiral stationary phases
Tachibana K and Ohnishi A
Journal of Chromatography A, 906(1-2), 127-154 (2001)
Thin-layer chromatography in food and agricultural analysis
Sherma J
Journal of Chromatography A, 880(1-2), 129-147 (2000)
Feasibility study on power ultrasound for regeneration of silica gel?a potential desiccant used in air-conditioning system
Yao Y, et al.
Applied Energy, 86(11), 2394-2400 (2009)
Convective drying of agitated silica gel and beech wood particle beds?experiments and transient DEM-CFD simulations
Sudbrock F, et al.
Drying Technology, 33(15-16), 1808-1820 (2015)
Thin-layer chromatography: a laboratory handbook
Thin Layer Chromatography: Reagents and Detection Methods (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service