Skip to Content
Merck
All Photos(1)

Documents

V800350

Sigma-Aldrich

Salicylic acid

LR, ≥99%

Synonym(s):

2-Hydroxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CO2H
CAS Number:
Molecular Weight:
138.12
Beilstein:
774890
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

LR

vapor density

4.8 (vs air)

vapor pressure

1 mmHg ( 114 °C)

product line

Vetec

Assay

≥99%

form

powder

bp

211 °C (lit.)

mp

158-161 °C (lit.)

density

1.44 g/cm3 at 20 °C

SMILES string

OC(=O)c1ccccc1O

InChI

1S/C7H6O3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H,(H,9,10)

InChI key

YGSDEFSMJLZEOE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

314.6 °F - closed cup

Flash Point(C)

157 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shifeng Cao et al.
Food chemistry, 134(4), 1715-1718 (2013-02-28)
The effect of elicitors associated with host defence on betacyanin accumulation in Amaranthus mangostanus seedlings was investigated. Under the conditions of the experiments, betacyanin accumulation was generally enhanced by light. Methyl jasmonate (MeJA) treatment increased betacyanin synthesis in a concentration-dependent
Ganesan Karthikeyan et al.
British journal of haematology, 146(2), 142-149 (2009-05-15)
Guidelines differ on whether acetyl salicylic acid (ASA, aspirin) should be used for prophylaxis in patients at high-risk of venous thromboembolism (VTE), principally because of differences in perceptions of its efficacy. ASA is an attractive therapeutic option because it is
A Corina Vlot et al.
Annual review of phytopathology, 47, 177-206 (2009-04-30)
For more than 200 years, the plant hormone salicylic acid (SA) has been studied for its medicinal use in humans. However, its extensive signaling role in plants, particularly in defense against pathogens, has only become evident during the past 20
E Benfeldt et al.
Acta dermato-venereologica, 79(5), 338-342 (1999-09-24)
Our aim was to simultaneously investigate 2 techniques for in vivo sampling of peripheral compartment pharmacokinetics after systemic administration of acetylsalicylic acid. Ten volunteers were given 2 g acetylsalicylic acid orally. Blood samples and dialysates from 4 microdialysis probes inserted
S Zaugg et al.
Journal of chromatography. B, Biomedical sciences and applications, 752(1), 17-31 (2001-03-20)
Acetylsalicylic acid (Aspirin) is rapidly metabolized to salicylic acid (salicylate) and other compounds, including gentisic acid and salicyluric acid. Monitoring of salicylate and its metabolites is of toxicological, pharmacological and biomedical interest. Three capillary electrophoresis (CE) methods featuring alkaline aqueous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service