Skip to Content
Merck
All Photos(1)

Key Documents

8.00691

Sigma-Aldrich

5-Sulfosalicylic acid dihydrate

for synthesis

Synonym(s):

5-Sulfosalicylic acid dihydrate, Salicylsulfonic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€42.20
250 G
€45.00
1 KG
€110.00

€42.20


Estimated to ship on29 March 2025


Request a Bulk Order

Select a Size

Change View
100 G
€42.20
250 G
€45.00
1 KG
€110.00

About This Item

Empirical Formula (Hill Notation):
C7H6O6S · 2H2O
CAS Number:
Molecular Weight:
254.21
MDL number:
UNSPSC Code:
12352106
EC Index Number:
202-555-6
NACRES:
NA.22
pH:
<0.5 (20 °C, 200 g/L in H2O)

€42.20


Estimated to ship on29 March 2025


Request a Bulk Order

vapor pressure

<1 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (acidimetric)

form

crystals

potency

1850 mg/kg LD50, oral (Rat)

pH

<0.5 (20 °C, 200 g/L in H2O)

mp

109-117 °C (anhydrous substance)

transition temp

flash point 150 °C

solubility

soluble 987 g/L

density

0.8 g/cm3 at 20 °C

bulk density

310 kg/m3

storage temp.

2-30°C

SMILES string

[S](=O)(=O)(O)c1cc(c(cc1)O)C(=O)O.C.C

InChI

1S/C7H6O6S.2CH4/c8-6-2-1-4(14(11,12)13)3-5(6)7(9)10;;/h1-3,8H,(H,9,10)(H,11,12,13);2*1H4

InChI key

NFYHZVWMQHQKRU-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 99,0 %(m)
Heavy metals (as Pb): ≤ 0,001 %(m)
Identity (IR): conforms

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service