Skip to Content
Merck
All Photos(1)

Key Documents

V800324

Sigma-Aldrich

Potassium iodate

AR, ≥99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KIO3
CAS Number:
Molecular Weight:
214.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥99.9%
grade:
AR
form:
crystalline powder
Pricing and availability is not currently available.

grade

AR

product line

Vetec

Assay

≥99.9%

form

crystalline powder

pH

5.5-9.0 (5% in water)

mp

560 °C (lit.)

density

3.93 g/mL at 25 °C (lit.)

cation traces

Fe: ≤0.001%
heavy metals (as Pb): ≤0.001%

SMILES string

[K+].[O-]I(=O)=O

InChI

1S/HIO3.K/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

JLKDVMWYMMLWTI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabian Rohner et al.
Food and nutrition bulletin, 33(4 Suppl), S330-S335 (2013-03-01)
Despite considerable progress made in the past decade through salt iodization programs, over 2 billion people worldwide still have inadequate iodine intake, with devastating consequences for brain development and intellectual capacity. To optimize these programs with regard to salt iodine
Ralf Biebinger et al.
The British journal of nutrition, 102(9), 1362-1369 (2009-08-06)
Adverse sensory changes prevent the addition of highly bioavailable ferrous sulfate (FeSO4) to most wheat flours. Poorly absorbable reduced Fe powders are commonly used. Encapsulation of FeSO4 can overcome these sensory changes, but the particle size of commercial compounds is
C Dryden et al.
Journal of perinatology : official journal of the California Perinatal Association, 27(5), 320-322 (2007-04-25)
Congenital thyrotoxicosis is a rare and potentially fatal illness. We report a case in a preterm infant delivered to a mother known to have autoimmune endocrine disease. Diagnosis was difficult because the infant's presenting symptoms and signs closely resembled congenital
Qiang Ren et al.
Journal of trace elements in medicine and biology : organ of the Society for Minerals and Trace Elements (GMS), 22(1), 1-8 (2008-03-06)
Iodine deficiency disorders continue to be a severe problem in many parts of Central Asia, causing delayed mental development and cretinism in indigenous populations. In some areas, iodized salt has not succeeded in controlling this problem. In southern Xinjiang Province
Mohsen Zeeb et al.
Talanta, 78(2), 584-589 (2009-02-11)
In this work, an on-line system with vapor-phase generation (VPG) and Fourier transform infrared (FTIR) spectrometric detection has been developed as a direct and highly selective analytical technique for the assay of penicillamine (PA). Potassium iodate solution was injected into

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service