Skip to Content
Merck
All Photos(1)

Key Documents

V800321

Sigma-Aldrich

Potassium hydroxide

LR, flakes, ≥85%

Synonym(s):

Caustic potash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOH
CAS Number:
Molecular Weight:
56.11
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
grade:
LR
form:
flakes
solubility:
H2O: soluble 1120 g/L

grade

LR

vapor pressure

1 mmHg ( 719 °C)

product line

Vetec

Assay

≥85%

form

flakes

pH

~13.5 (25 °C, 5.6 g/L)

mp

361 °C (lit.)

solubility

H2O: soluble 1120 g/L

anion traces

carbonate (CO32-): ≤2.0%

SMILES string

[OH-].[K+]

InChI

1S/K.H2O/h;1H2/q+1;/p-1

InChI key

KWYUFKZDYYNOTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashok R Patel et al.
Journal of agricultural and food chemistry, 63(19), 4862-4869 (2015-05-02)
The aim of this study was to use a detailed rheological characterization to gain new insights into the gelation behavior of natural waxes. To make a comprehensive case, six natural waxes (differing in the relative proportion of chemical components: hydrocarbons
Mizuki Takemoto et al.
PloS one, 10(6), e0131094-e0131094 (2015-06-27)
Channelrhodopsin (ChR) is a light-gated cation channel that responds to blue light. Since ChR can be readily expressed in specific neurons to precisely control their activities by light, it has become a powerful tool in neuroscience. Although the recently solved
Peng Li et al.
Angewandte Chemie (International ed. in English), 54(2), 574-577 (2014-11-15)
Self-assembly of a trigonal building subunit with diaminotriazines (DAT) functional groups leads to a unique rod-packing 3D microporous hydrogen-bonded organic framework (HOF-3). This material shows permanent porosity and demonstrates highly selective separation of C2H2/CO2 at ambient temperature and pressure.
Marilia Freitas Calmon et al.
Colloids and surfaces. B, Biointerfaces, 100, 177-184 (2012-07-07)
Magnetic iron oxide nanoparticles (magnetite) (MNPs) were prepared using different organic and inorganic bases. Strong inorganic base (KOH) and organic bases (NH(4)OH and 1,4-diazabicyclo[2.2.2]octane (DABCO)) were used in the syntheses of the MNPs. The MNPs were characterized by X-ray diffraction
K Y Foo et al.
Bioresource technology, 130, 696-702 (2013-01-22)
In this work, preparation of granular activated carbon from oil palm biodiesel solid residue, oil palm shell (PSAC) by microwave assisted KOH activation has been attempted. The physical and chemical properties of PSAC were characterized using scanning electron microscopy, volumetric

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service