Skip to Content
Merck
All Photos(2)

Key Documents

V800126

Sigma-Aldrich

Cobalt(II) nitrate hexahydrate

LR, ≥97%

Synonym(s):

Cobaltous nitrate, Cobaltous nitrate hexahydrate, Nitric acid, cobalt(II) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Co(NO3)2 · 6H2O
CAS Number:
Molecular Weight:
291.03
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
LR
form:
crystals

grade

LR

product line

Vetec

Assay

≥97%

form

crystals

mp

55 °C (lit.)

anion traces

chloride (Cl-): ≤0.005%

SMILES string

O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Co.2NO3.6H2O/c;2*2-1(3)4;;;;;;/h;;;6*1H2/q+2;2*-1;;;;;;

InChI key

QGUAJWGNOXCYJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Dam. 1 - Muta. 2 - Ox. Sol. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 2 Inhalation

Target Organs

Lungs

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William Buchmann et al.
Journal of mass spectrometry : JMS, 42(4), 517-526 (2007-02-27)
The electronebulization of a cobalt(II)/cysteine(Cys) mixture in water/methanol (50/50) produced mainly cobalt-cationized species. Three main groups of the Co-cationized species can be distinguished in the ESI-MS spectrum: (1) the cobalt complexes including the cysteine amino acid only (they can be
Massimiliano Aschi et al.
Journal of mass spectrometry : JMS, 44(7), 1038-1046 (2009-04-01)
The nature of the ionic species, formed by electrospray ionization (ESI) of Co(NO3)2/CH3OH solutions with a pair of aminoalcohols W and Y, has been investigated by mass spectrometric and computational methods. Collision induced dissociation (CID) of ions, formally corresponding to
H H Chen et al.
Nucleic acids research, 10(11), 3561-3571 (1982-06-11)
Transient electric dichroism measurements have been used to observe the rotational relaxation times of 145 base pair fragments of poly (dGm5dC) and random sequence DNA to solution. From these the lengths of the fragments are calculated and the interbase pair
N G Al-Tawil et al.
Acta dermato-venereologica, 64(3), 203-208 (1984-01-01)
Peripheral blood lymphocytes from 45 patients with a positive patch test to cobalt chloride (CoCl2) and 37 controls were stimulated with various concentrations of CoCl2 and/or cobalt sulphate (CoSO4) or cobalt nitrate (Co(NO3)2) or cobalt acetate [CH3COO)2Co) for various days
A time-resolved in situ quick-XAS investigation of thermal activation of Fischer-Tropsch silica-supported cobalt catalysts.
Jingping Hong et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(10), 2802-2805 (2012-02-07)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service