Skip to Content
Merck
All Photos(1)

Key Documents

63539

Sigma-Aldrich

Manganese(II) carbonate hydrate

44-46% Mn basis (KT)

Synonym(s):

Manganese(2+) carbonate hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€36.40
1 KG
€103.00

€36.40


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
250 G
€36.40
1 KG
€103.00

About This Item

Linear Formula:
MnCO3 · xH2O
CAS Number:
Molecular Weight:
114.95 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

€36.40


Estimated to ship on31 May 2025


Request a Bulk Order

grade

for analytical purposes

form

powder

concentration

44-46% Mn (KT)

SMILES string

[Mn++].[O-]C([O-])=O

InChI

1S/CH2O3.Mn/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

XMWCXZJXESXBBY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service