Skip to Content
Merck
All Photos(1)

Key Documents

V800081

Sigma-Aldrich

Benzyl chloride

AR, ≥99%

Synonym(s):

α-Chlorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2Cl
CAS Number:
Molecular Weight:
126.58
Beilstein:
471308
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

AR

vapor density

4.36 (vs air)

vapor pressure

10.3 mmHg ( 60 °C)
7 mmHg ( 55 °C)

product line

Vetec

Assay

≥99%

autoignition temp.

1085 °F

expl. lim.

14 %

refractive index

n20/D 1.538 (lit.)

bp

177-181 °C (lit.)

mp

−43 °C (lit.)

density

1.1 g/mL at 25 °C (lit.)

functional group

chloro
phenyl

SMILES string

ClCc1ccccc1

InChI

1S/C7H7Cl/c8-6-7-4-2-1-3-5-7/h1-5H,6H2

InChI key

KCXMKQUNVWSEMD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Heart,forestomach, Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P B Nagabalasubramanian et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 77(1), 150-159 (2010-06-12)
The FT-IR and FT-Raman vibrational spectra of alpha-chlorotoluene have been recorded using Perkin-Elmer 180 spectrometer in the range 3600-10 cm(-1) in the solid phase. A detailed vibrational spectral analysis has been carried out and assignments of the observed fundamental bands
Muriel Amatore et al.
Chemical communications (Cambridge, England), (40)(40), 5019-5021 (2008-10-22)
A new cobalt-catalysed reductive coupling of aryl halides with benzyl chlorides is reported; a variety of diarylmethanes can be prepared in good to excellent yields under mild reaction conditions using CoBr(2) as catalyst and Zn dust; this new cobalt-catalysed coupling
Ryosuke Matsubara et al.
Journal of the American Chemical Society, 133(47), 19020-19023 (2011-11-10)
Nickel-catalyzed intermolecular benzylation and heterobenzylation of unactivated alkenes to provide functionalized allylbenzene derivatives are described. A wide range of both the benzyl chloride and alkene coupling partners are tolerated. In contrast to analogous palladium-catalyzed variants of this process, all reactions
Oxidatively intercepting Heck intermediates: Pd-catalyzed 1,2- and 1,1-arylhalogenation of alkenes.
Dipannita Kalyani et al.
Journal of the American Chemical Society, 130(7), 2150-2151 (2008-01-31)
Mikael P Johansson et al.
Dalton transactions (Cambridge, England : 2003), 40(33), 8419-8428 (2011-07-21)
α-diimine iron complexes have been suggested to catalyse polymerisation via two distinct pathways, depending on the spin state of the iron complex. Here, we study a typical complex of this family, (R'')[N,N]FeCl(2), with [N,N] = Cy-N=CR''-CR''=N-Cy (Cy = cyclohexyl, R''

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service