Skip to Content
Merck
All Photos(1)

Documents

8.01809

Sigma-Aldrich

Benzyl chloride

for synthesis

Synonym(s):

Benzyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7Cl
CAS Number:
Molecular Weight:
126.58
MDL number:
UNSPSC Code:
12352101
EC Index Number:
202-853-6
NACRES:
NA.22

vapor pressure

1.2 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

585 °C

potency

440 mg/kg LD50, oral (Rat)

expl. lim.

1.1-14 % (v/v)

bp

179.3 °C/1013 hPa

mp

-39 °C

transition temp

flash point 67 °C

solubility

460 mg/L

density

1.10 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C7H7Cl/c8-6-7-4-2-1-3-5-7/h1-5H,6H2

InChI key

KCXMKQUNVWSEMD-UHFFFAOYSA-N

Application

  • Research on the Oxidative Chlorination of Toluene to Benzyl Chloride by Chloride Ion: This study explores the preparation of benzyl chloride through oxidative chlorination under LED blue light, achieving a 40% yield (Y Bai, X Cheng, A Zheng, H Shen, 2019).
  • Tertiary amines nucleophilicity in quaternization reaction with benzyl chloride: Investigates the kinetics of the quaternization reaction of tertiary amines with benzyl chloride, providing insights into reaction mechanisms and conditions (KS Yutilova, SG Bakhtin, OM Shved, et al., 2015).
  • Synthesis and inhibition profiles of N-benzyl-and N-allyl aniline derivatives against carbonic anhydrase and acetylcholinesterase–A molecular docking study: Focuses on the synthesis of N-benzyl derivatives and their biological activity, utilizing benzyl chloride as a key intermediate in the synthesis process (I Mahmudov, Y Demir, Y Sert, Y Abdullayev, et al., 2022).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.099 - 1.100
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

152.6 °F

Flash Point(C)

67 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service